Skip to Content
Merck
All Photos(3)

Key Documents

HPA040825

Sigma-Aldrich

Anti-IP6K1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-IHPK1, Anti-Inositol hexakisphosphate kinase 1, Anti-KIAA0263

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL

immunogen sequence

DSDGYINLVAYPYVESETVEQDDTTEREQPRRKHSRRSLHRSGSGSDHKEEKASLSLETSESSQEAKSP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IP6K1(9807)

General description

Inositol hexakisphosphate kinase (IP6K1) is encoded by the gene mapped to human chromosome 3p21.31. The encoded protein belongs to the family of inositol hexakisphosphate (IP6) proteins and is widely expressed.

Immunogen

inositol hexakisphosphate kinase 1 recombinant protein epitope signature tag (PrEST)

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.

Biochem/physiol Actions

Inositol hexakisphosphate kinase 1 (IP6K1) is involved in the synthesis of diphosphoinositol pentakisphosphate. It interacts with cullin–RING ubiquitin ligase-4 (CRL4) and has a role in DNA homologous recombination repair. In addition, IP6K1 is also implicated in chromatin modifications and spermatogenesis. Elevated expression or mutation of IP6K1 is associated with the pathogenesis of type 2 diabetes mellitus and crohn′s disease. IP6K1 plays a vital role in dynein-dependent trafficking pathways, including endosomal sorting, vesicle movement and Golgi maintenance. IP6K1 controls DNA repair and cell death by mediating disassembly of the cullin-RING ubiquitin ligase 4 (CRL4)-COP9 signalosome (CSN) complex.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST79496

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Contribution of polymorphic variation of inositol hexakisphosphate kinase 3 (IP6K3) gene promoter to the susceptibility to late onset Alzheimer's disease.
Crocco P
Biochimica et Biophysica Acta, 1862(9), 1766-1773 (2016)
Characterization of a selective inhibitor of inositol hexakisphosphate kinases: use in defining biological roles and metabolic relationships of inositol pyrophosphates.
Padmanabhan U
The Journal of Biological Chemistry, 284(16), 10571-10582 (2009)
Masatoshi Ito et al.
The Journal of biological chemistry, 299(3), 102928-102928 (2023-01-22)
Inositol pyrophosphates regulate diverse physiological processes; to better understand their functional roles, assessing their tissue-specific distribution is important. Here, we profiled inositol pyrophosphate levels in mammalian organs using an originally designed liquid chromatography-mass spectrometry (LC-MS) protocol and discovered that the
Chunfang Gu et al.
PloS one, 11(10), e0165286-e0165286 (2016-10-28)
The HCT116 cell line, which has a pseudo-diploid karotype, is a popular model in the fields of cancer cell biology, intestinal immunity, and inflammation. In the current study, we describe two batches of diverged HCT116 cells, which we designate as
Rathan S Jadav et al.
Cellular signalling, 28(8), 1124-1136 (2016-05-04)
Inositol hexakisphosphate kinases (IP6Ks), a family of enzymes found in all eukaryotes, are responsible for the synthesis of 5-diphosphoinositol pentakisphosphate (5-IP7) from inositol hexakisphosphate (IP6). Three isoforms of IP6Ks are found in mammals, and gene deletions of each isoform lead

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service