Skip to Content
Merck
All Photos(8)

Key Documents

HPA037470

Sigma-Aldrich

Anti-DNAH5 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CILD3, Anti-Dnahc5, Anti-Dynein, axonemal, heavy chain 5, Anti-HL1, Anti-KTGNR, Anti-PCD

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:500-1:1000

immunogen sequence

EVEDAILEGNQIERIDQLFAVGGLRHLMFYYQDVEEAETGQLGSLGGVNLVSGKIKKPKVFVTEGNDVALTGVCVFFIRTDPSKAITPD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DNAH5(1767)

General description

The gene DNAH5 (dynein axonemal heavy chain 5) is mapped to human chromosome 5p15.3. It is a component of the microtubule-linked motor protein complex.

Immunogen

dynein, axonemal, heavy chain 5 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies®Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-DNAH5 antibody produced in rabbit has been used in western blotting and immunofluorescence.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

DNAH5 (dynein axonemal heavy chain 5) is an axonemal heavy chain dynein protein. With the help of the ATPase activity, this protein works as a force generator in respiratory cilia. Mutation in this gene is linked with primary ciliary dyskinesia (PCD) type 3 and Kartagener syndrome. In chronic obstructive pulmonary disease, DNAH5 is associated with total lung capacity and regulates hyperinflation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST79923

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Targeted NGS gene panel identifies mutations in RSPH1 causing primary ciliary dyskinesia and a common mechanism for ciliary central pair agenesis due to radial spoke defects.
Onoufriadis A, et al.
Human Molecular Genetics, 23, 3362-3374 (2014)
Application of SNP microarrays to the genome-wide analysis of chromosomal instability in premalignant airway lesions.
Nakachi I, et al.
Cancer Prevention Research (Philadelphia, Pa.), 7, 255-265 (2014)
Combined exome and whole-genome sequencing identifies mutations in ARMC4 as a cause of primary ciliary dyskinesia with defects in the outer dynein arm.
Onoufriadis A, et al.
Journal of medical Genetics, 51, 61-67 (2014)
HEATR2 plays a conserved role in assembly of the ciliary motile apparatus.
Diggle CP, et al.
PLoS Genetics, 10, e1004577-e1004577 (2014)
Zakia A Abdelhamed et al.
International journal of gynecological cancer : official journal of the International Gynecological Cancer Society, 28(8), 1535-1544 (2018-08-11)
The aim of this study was to investigate the distribution of primary cilia on secretory cells in normal fallopian tube (FT) and serous tubal intraepithelial carcinoma (STIC). Fallopian tube tissue samples were obtained from 4 females undergoing prophylactic hysterectomies and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service