Skip to Content
Merck
All Photos(7)

Documents

HPA031345

Sigma-Aldrich

Anti-SLC2A1 antibody produced in rabbit

enhanced validation

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-DYT18, Anti-GLUT, Anti-GLUT1, Anti-HTLVR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

GRTFDEIASGFRQGGASQSDKTPEELFHP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC2A1(6513)

General description

The SLC2A1 (solute carrier family 2 member 1) gene is 35kb in length and is mapped to human chromosome 1p35 → p31.3. The gene contains 10 exons and nine introns. Its mRNA is found to be expressed in several tissues. It is highly expressed in cerebral endothelial cells and the encoded protein has 12 transmembrane domains.

Immunogen

solute carrier family 2 (facilitated glucose transporter), member 1

Application

Anti-SLC2A1 antibody produced in rabbit has been used in immunohistochemistry.

Biochem/physiol Actions

The SLC2A1 (solute carrier family 2 member 1) gene, also referred to as GLUT1, encodes protein that is mainly involved in the transport of D-glucose across the blood-brain barrier. Mutation in this gene causes Glut-1 deficiency syndrome, which is characterized by reduced cerebrospinal fluid glucose concentrations (hypoglycorrhachia) and reduced erythrocyte glucose transporter activities in the patients along with infantile seizures, acquired microcephaly and developmental delay. It has been found to be critical in the development of the blood-brain barrier. Mutation in this gene has also been associated with paroxysmal exercise-induced dyskinesia and epilepsy.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST87043

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chiara Levra Levron et al.
Nature cell biology, 25(5), 740-753 (2023-04-21)
Epithelial cells that participated in wound repair elicit a more efficient response to future injuries, which is believed to be locally restricted. Here we show that cell adaptation resulting from a localized tissue damage has a wide spatial impact at
Yang Chen et al.
Heliyon, 10(12), e33077-e33077 (2024-07-12)
Dysfunction of the blood-brain barrier (BBB) has been increasingly recognised as a critical early event in Alzheimer's disease (AD) pathophysiology. Central to this mechanism is the impaired function of brain endothelial cells (BECs), the primary structural constituents of the BBB
Mutational analysis of GLUT1 (SLC2A1) in Glut-1 deficiency syndrome.
Wang D, et al.
Human Genetics, 16(3), 224-231 (2000)
Qi Wang et al.
Brain : a journal of neurology, 145(12), 4474-4488 (2022-07-06)
Alzheimer's disease is a neurodegenerative disorder that causes age-dependent neurological and cognitive declines. The treatments for Alzheimer's disease pose a significant challenge, because the mechanisms of disease are not being fully understood. Malfunction of the blood-brain barrier is increasingly recognized
Xiaomin Huang et al.
Cell death & disease, 15(4), 258-258 (2024-04-13)
The impairment of the blood-brain barrier (BBB) has been increasingly recognised as a critical element in the early pathogenesis of Alzheimer's disease (AD), prompting a focus on brain endothelial cells (BECs), which serve as the primary constituents of the BBB.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service