Skip to Content
Merck
All Photos(4)

Documents

HPA029590

Sigma-Aldrich

Anti-SLC11A1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Lsh, Anti-Nramp, Anti-Nramp1, Anti-Solute carrier family 11 (proton-coupled divalent metal ion transporters), member 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

TSPGPQQAPPRETYLSEKIPIPDTKPGTFSLR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC11A1(6556)

General description

Solute carrier family 11 member 1 (SLC11A1) is a multi-pass membrane protein. It is expressed in macrophages and neutrophils. The gene is located on human chromosome 2q35.

Immunogen

solute carrier family 11 (proton-coupled divalent metal ion transporters), member 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Solute carrier family 11 member 1 (SLC11A1) helps in transporting divalent metal ions (Mn2+ and Fe2+) and iron metabolism. It contributes to host innate immune response against infections. The protein regulates CXC chemokine KC, interleukin-1b (IL-1b), inducible nitric oxide synthase (iNOS), major histocompatibility complex (MHC) class II molecules, tumor necrosis factor α (TNFα), nitric oxide (NO) release, L-arginine flux, oxidative burst, tumoricidal and antimicrobial activity. The protein plays an important role in disease pathology and susceptibility. Mutations in this gene is linked to rheumatoid arthritis, tuberculosis and inflammatory bowel disease.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST78903

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

SLC11A1 polymorphisms and host susceptibility to cutaneous leishmaniasis in Pakistan
Sophie M, et al.
Parasites & vectors, 10(1), 12-12 (2017)
SLC11A1 (formerly NRAMP1) and disease resistance: Microreview
Blackwell JM, et al.
Cellular Microbiology, 3(12), 773-784 (2001)
Twenty-four signature genes predict the prognosis of oral squamous cell carcinoma with high accuracy and repeatability
Gao J, et al.
Molecular Medicine Reports, 17(2), 2982-2990 (2018)
Genetic variants of SLC11A1 are associated with both autoimmune and infectious diseases: systematic review and meta-analysis
Archer NS, et al.
Genes and Immunity, 16(4), 275-275 (2015)
Genomewide linkage analysis of the granulomatous mitsuda reaction implicates chromosomal regions 2q35 and 17q21
Ranque B, et al.
The Journal of Infectious Diseases, 196(8), 1248-1252 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service