Skip to Content
Merck
All Photos(4)

Key Documents

HPA025813

Sigma-Aldrich

Anti-XRCC5 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-86 kDa subunit of Ku antigen, Anti-ATP-dependent DNA helicase 2 subunit 2, Anti-ATP-dependent DNA helicase II 80 kDa subunit, Anti-CTC85, Anti-CTCBF, Anti-DNA repair protein XRCC5, Anti-Ku80, Anti-Ku86, Anti-Lupus Ku autoantigen protein p86, Anti-Nuclear factor IV, Anti-TLAA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

LKEDIIQGFRYGSDIVPFSKVDEEQMKYKSEGKCFSVLGFCKSSQVQRRFFMGNQVLKVFAARDDEAAAVALSSLIHALDDLDMVAIVRYAYDKRANPQVGVAFPHIKHNYECLVYVQLPFMEDLRQYMFSSLKNSKKYAPTE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... XRCC5(7520)

Immunogen

ATP-dependent DNA helicase 2 subunit 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The gene XRCC5 (X-ray repair cross complementing 5) is mapped to human chromosome 2q35. It is part of the non-homologous end joining double-strand break repair pathway. XRCC5 helps in damage recognition. Mutations in the gene are associated with risk of glioma and chronic obstructive pulmonary disease (COPD). Mutation in it might also be linked with gastric cancer development, particularly in cases of family history. XRCC5 is also involved in alcohol dependence. In addition, it is a part of the OCFRE (osteocalcin (OC) fibroblast growth factor (FGF) response element (FRE))-binding complex, thereby controlling the gene expression of osteocalcin.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71055

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regional assignment of a human DNA repair gene (XRCC5) to 2q35 by X-ray hybrid mapping.
Chen DJ, et al.
Genomics, 21, 423-427 (1994)
Regulation of osteocalcin gene expression by a novel Ku antigen transcription factor complex.
Willis DM, et al.
The Journal of Biological Chemistry, 277, 37280-37291 (2002)
XRCC5 as a risk gene for alcohol dependence: evidence from a genome-wide gene-set-based analysis and follow-up studies in Drosophila and humans.
Juraeva D, et al.
Neuropsychopharmacology, 40, 361-371 (2015)
Susceptibility to gastric cancer and polymorphisms of insertion/deletion at the intron 3 of the XRCC4 and VNTR at the promoter region of the XRCC5.
Saadat M, et al.
Pathology Oncology Research, 21, 689-693 (2015)
Association of XRCC5 polymorphisms with COPD and COPD-related phenotypes in the Han Chinese population: a case-control cohort study.
Wang B, et al.
Genetics and molecular research : GMR, 13, 7070-7078 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service