Skip to Content
Merck
All Photos(4)

Key Documents

HPA025235

Sigma-Aldrich

Anti-VANGL1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-LPP2, Anti-Loop-tail protein 2 homolog, Anti-Strabismus 2, Anti-Van Gogh-like protein 1, Anti-Vang-like protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
RNAi knockdown
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL

immunogen sequence

DTESTYSGYSYYSSHSKKSHRQGERTRERHKSPRNKDGRGSEKSVTIQPPTGEPLLGNDSTRTEEVQDDNWGETTTAITGTSEHSISQEDIARISKDMEDSVGLDCKRY

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VANGL1(81839)

General description

The gene VANGL1 (vang-like protein 1) is mapped to human chromosome 1p13. The encoded protein is a transmembrane protein and a component of the Wnt-PCP (planar cell polarity) pathway. The VANGL1 protein can interact with planar cell polarity (PCP) core proteins Disheveled, Prickle, and Frizzled, KAI1 (metastasis suppressor Kangai-1) protein and ITF (intestinal trefoil factor) protein.

Immunogen

Vang-like protein 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies®Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-VANGL1 antibody produced in rabbit has been used for immunohistochemsitry and immunofluorescence.
Anti-VANGL1 antibody produced in rabbit has been used in western blot and immunostaining.

Biochem/physiol Actions

VANGL1 (vang-like protein 1) is associated with tumor progression in various cancers. In colorectal cancer, it enhances the angiogenesis. In mouse colon cells, overexpression of the VANGL1 gene causes tumorigenicity, invasiveness and adhesion to fibronectin. VANGL1 is upregulated in colon, laryngeal, oral cavity squamous, gastric and hepatocellular cancer tissues. VANGL1 is a planar cell polarity protein. It plays a significant role in embryogenesis and is required for normal embryonic development.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73163

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michelle L Stoller et al.
Developmental biology, 437(1), 17-26 (2018-03-07)
The organization of polarized stereociliary bundles is critical for the function of the inner ear sensory receptor hair cells that detect sound and motion, and these cells present a striking example of Planar Cell Polarity (PCP); the coordinated orientation of
A novel electrochemical immunosensor based on the rGO-TEPA-PTC-NH2 and AuPt modified C60 bimetallic nanoclusters for the detection of Vangl1, a potential biomarker for dysontogenesis.
Chen Q, et al.
Biosensors And Bioelectronics, 79, 364-370 (2016)
Gokhan Ozan Cetin et al.
Genetic testing and molecular biomarkers, 19(6), 283-287 (2015-04-16)
The Wnt planar cell polarity (PCP) pathway is one of the Wnt pathways which plays a critical role in cell proliferation and fate. The VANGL1 protein is one of Wnt-PCP pathway components. It is known that Wnt-PCP pathway has major
Renata Prunskaite-Hyyryläinen et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 28(4), 1568-1581 (2013-12-29)
Wnt4 is a key signal that channels the developmental fate of the indifferent mammalian gonad toward the ovary, but whether Wnt4 has later roles during ovary development remains unknown. To investigate this, we inactivated the Wnt4 gene by crossing Amhr2Cre
Katsura Minegishi et al.
Developmental cell, 40(5), 439-452 (2017-03-16)
Polarization of node cells along the anterior-posterior axis of mouse embryos is responsible for left-right symmetry breaking. How node cells become polarized has remained unknown, however. Wnt5a and Wnt5b are expressed posteriorly relative to the node, whereas genes for Sfrp

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service