Skip to Content
Merck
All Photos(8)

Key Documents

HPA023616

Sigma-Aldrich

Anti-CDH17 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-Cadherin-17, Anti-Intestinal peptide-associated transporter HPT-1, Anti-LI-cadherin, Anti-Liver-intestine cadherin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
recombinant expression
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

HPIKITQVRWNDPGAQYSLVDKEKLPRFPFSIDQEGDIYVTQPLDREEKDAYVFYAVAKDEYGKPLSYPLEIHVKVK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDH17(1015)

General description

Cadherin-17 is a liver-intestine cadherin protein encoded by the CDH17 gene in humans and belongs to unique member of cadherin superfamily. CDH17 is a non-classical cadherin having seven cadherin domains and a transmembrane domain but lacking the conserved cytoplasmic domain of classical cadherins. Cadherins are cellular adhesion molecules that are crucial for intracellular adhesion. These are considered as potential tumor suppressor genes and any alterations in them may lead to invasion and metastasis of tumor cells. The gene CDH17 is mapped to human chromosome 8q22.1 and contains 18 exons.

Immunogen

Cadherin-17 Precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-CDH17 antibody produced in rabbit has been used in immunohistochemistry.
All Prestige Antibodies®Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The protein encoded by the gene CDH17 (Cadherin-17), also referred to as LI-cadherin, functions in cell adhesion without being dependent on cytoplasmic components, such as catenins or the actin cytoskeleton. Lower expression of this protein due to single nucleotide polymorphisms has been associated with tumor progression and lymph node metastasis of human colorectal carcinoma. It is found to regulate the signaling function of α2β1 integrin in cell adhesion and proliferation in liver metastatic cancer cells.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76191

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kristina Magnusson et al.
The American journal of surgical pathology, 35(7), 937-948 (2011-06-17)
The special AT-rich sequence-binding protein 2 (SATB2), a nuclear matrix-associated transcription factor and epigenetic regulator, was identified as a tissue type-specific protein when screening protein expression patterns in human normal and cancer tissues using an antibody-based proteomics approach. In this
Tingting Bian et al.
Oncotarget, 8(38), 63442-63452 (2017-10-04)
Pulmonary enteric adenocarcinoma (PEAC), a rare type of non-small cell lung cancer, has similar histological and immunohistochemical morphology to colorectal adenocarcinoma. Cadherin-17 (CDH17) and SATB homeobox 2 (SATB2) immunoexpression have recently been demonstrated in colorectal adenocarcinoma. In this study, we
Single nucleotide polymorphisms in the CDH17 gene of colorectal carcinoma.
Chen RY
World Journal of Gastroenterology, 18, 7251-7261 (2012)
Alexandra K Eicher et al.
Cell stem cell, 29(1), 36-51 (2021-12-03)
Human organoid model systems lack important cell types that, in the embryo, are incorporated into organ tissues during development. We developed an organoid assembly approach starting with cells from the three primary germ layers-enteric neuroglial, mesenchymal, and epithelial precursors-that were
Jin Wang et al.
Cancer biology & therapy, 14(3), 262-270 (2013-01-10)
Cadherin-17 (CDH17), as a structurally unique member of the cadherin superfamily, has been identified to predict a poor prognosis for gastric cancer (GC). Our previous study demonstrated the positive correlation between CDH17 and lymph node micrometastasis in GC. We sought

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service