Skip to Content
Merck
All Photos(1)

Key Documents

HPA023277

Sigma-Aldrich

Anti-SEMA4D antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Putative uncharacterized protein C9orf164

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

RCSVLSSAGNKTSKVQVAVMRPEVTHQERWTRELSAWRAVAGEHDRMMQSWRKAWESCSKDTL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SEMA4D(10507)

General description

The gene SEMA4D (semaphorin 4D) is mapped to human chromosome 9q22. SEMA4D transcripts are mainly expressed in the skeletal muscle, peripheral blood lymphocytes, spleen and thymus. However, low expression of SEMA4D can be seen in the testes, brain, kidney, small intestine, prostate, heart, placenta, lung and pancreas. In T cells, SEMA4D localizes at the membrane and is also present in the soluble form. It is a member of the class four of the semaphorin protein family.

Immunogen

Putative uncharacterized protein C9orf164 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SEMA4D (semaphorin 4D) protein is mainly involved in the nervous and immune systems. It regulates migration and cytokine generation in monocytes, participates in T cell activation, B cell survival, cell-to-cell communication and adhesion. SEMA4D plays a crucial role in atherogenesis and thrombus formation by helping in platelet-endothelial cell association. It is associated with breast cancer, colorectal carcinoma, cervical cancer, non-small cell lung cancer, rheumatoid arthritis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75410

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jin-Shen Wang et al.
World journal of gastroenterology, 21(7), 2191-2198 (2015-02-27)
To investigate semaphorin 4D (Sema4D) and hypoxia-inducible factor-1α (HIF-1α) expression in colorectal carcinoma and evaluate their clinicopathological and prognostic significance. Eighty-six curatively resected colorectal carcinoma patients at different stages of disease were randomly selected from the group of patients who
Maria Carolina A Luque et al.
Molecular immunology, 67(2 Pt B), 559-567 (2015-08-16)
Leukocyte migration is essential for the function of the immune system. Their recruitment from the vessels to the tissues involves sequential molecular interactions between leukocytes and endothelial cells (ECs). Many adhesion molecules involved in this process have already been described.
Yuji Yoshida et al.
Arthritis & rheumatology (Hoboken, N.J.), 67(6), 1481-1490 (2015-02-25)
Semaphorin 4D (Sema4D)/CD100 has pleiotropic roles in immune activation, angiogenesis, bone metabolism, and neural development. We undertook this study to investigate the role of Sema4D in rheumatoid arthritis (RA). Soluble Sema4D (sSema4D) levels in serum and synovial fluid were analyzed
K T Hall et al.
Proceedings of the National Academy of Sciences of the United States of America, 93(21), 11780-11785 (1996-10-15)
Herein we describe the molecular characterization of the human leukocyte activation antigen CD100 and identify it as the first semaphorin, to our knowledge, in the immune system. Semaphorins have recently been described as neuronal chemorepellants that direct pioneering neurons during
Sha-sha Ruan et al.
Journal of Huazhong University of Science and Technology. Medical sciences = Hua zhong ke ji da xue xue bao. Yi xue Ying De wen ban = Huazhong keji daxue xuebao. Yixue Yingdewen ban, 34(4), 491-496 (2014-08-20)
This study aimed to explore Semaphrin4D (Sema4D) expression and clinical significance in non-small cell lung cancer (NSCLC), and to define the roles and mechanisms of Sema4D in regulating the malignant behaviors of A549 cells by small interfering RNA (siRNA). Firstly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service