Skip to Content
Merck
All Photos(8)

Key Documents

HPA019462

Sigma-Aldrich

Anti-DEFA6 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Defensin, alpha 6, Anti-Defensin-6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:5000-1:10000

immunogen sequence

PLQAEDDPLQAKAYEADAQEQRGANDQDFAVSFAEDASSSLRALGSTRAFTCHCRRSCYSTEYSYGTCTVMGINHRFC

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DEFA6(1671)

General description

The gene DEFA6 (defensin-6) is mapped to human chromosome 8p23.1. It belongs to the defensin family of proteins. DEFA6 is expressed by paneth cells in the intestine.

Immunogen

Defensin-6 Precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-DEFA6 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

DEFA6 (defensin-6) is an antimicrobial peptide, which is mainly involved in intestinal innate immunity. DEFA6-mediated formation of fibrils and nanonets are crucial for enclosing and entraping enteric bacterial pathogens. It is down-regulated in Crohn′s disease (CD). DEFA6 also increases HIV (human immunodeficiency virus) infection by promoting virus attachment on the target cells.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74710

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carissa M Lucero et al.
Clinical and vaccine immunology : CVI, 20(8), 1320-1328 (2013-06-28)
CXCL13 is a constitutively expressed chemokine that controls migration of immune cells to lymphoid follicles. Previously, we found CXCL13 mRNA levels increased in rhesus macaque spleen tissues during AIDS. This led us to examine the levels and locations of CXCL13
Atle van Beelen Granlund et al.
Scandinavian journal of gastroenterology, 46(11), 1316-1323 (2011-10-14)
To do a genome-wide gene expression study of active and inactive ulcerative colitis and Crohn's disease (inflammatory bowel disease--IBD) and examine the most differentially expressed genes. As the study showed an extreme upregulation of all regenerating islet-derived genes (REG proteins)
Gao Tan et al.
European journal of cell biology, 94(1), 60-66 (2014-12-01)
Human enteric α-defensins (HD5 and HD6), major antimicrobial peptides produced by Paneth cells in the intestine, play important roles in intestinal innate immunity. Since their expression is decreased in Crohn's disease (CD), with decreased expression being more pronounced in the
Atle van Beelen Granlund et al.
Cell and tissue research, 352(3), 639-646 (2013-03-23)
The regenerating islet-derived (REG) gene family encodes a group of proteins highly expressed in several human pathologies, many of which are associated with epithelial inflammation. All human family members, namely REG1A, REG1B, REG3A and REG4, are closely related in genomic
Marcin Duleba et al.
Nature protocols, 15(5), 1612-1627 (2020-04-03)
'Adult' or 'somatic' stem cells harbor an intrinsic ability to regenerate tissues. Heterogeneity of such stem cells along the gastrointestinal tract yields the known segmental specificity of this organ and may contribute to the pathology of certain enteric conditions. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service