Skip to Content
Merck
All Photos(8)

Key Documents

HPA017998

Sigma-Aldrich

Anti-FOXK1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-5164497

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

PHDPEFGSKLASVPEYRYSQSAPGSPVSAQPVIMAVPPRPSSLVAKPVAYMPASIVTSQQPAGHAIHVVQQAPTVTMVRVVTTSANSANGYILTSQGAAGGSHDAAGAAVLDLGSEARGLEEKPTIAFATIPAAGGVIQTVA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FOXK1(221937)

General description

Forkhead box protein K1 (FOXK1) is expressed in tissues of the brain, heart and lungs during the developmental stages. The gene encoding FOXK1 is located on chromosome 7p22.

Immunogen

forkhead box K1

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Forkhead box protein K1 (FOXK1) interacts with dishevelled protein (DVL) and transports it to the nucleus. This is important for the progress and positive regulation of the Wnt signaling pathway. Knockdown of FOXK1 leads to diminished cell proliferation rates. FOXK1 binds and regulates genes that are related to cell cycle like DHFR (Dihydrofolate reductase) and TYMS (Thymidylate synthase). It has been shown that its expression is heightened in colorectal cancers.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73352

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenqi Wang et al.
Developmental cell, 32(6), 707-718 (2015-03-26)
Dishevelled (DVL) proteins serve as crucial regulators that transduce canonical Wnt signals to the GSK3β-destruction complex, resulting in the stabilization of β-catenin. Emerging evidence underscores the nuclear functions of DVLs, which are critical for Wnt/β-catenin signaling. However, the mechanism underlying
Gavin D Grant et al.
Molecular biology of the cell, 23(16), 3079-3093 (2012-06-29)
We developed a system to monitor periodic luciferase activity from cell cycle-regulated promoters in synchronous cells. Reporters were driven by a minimal human E2F1 promoter with peak expression in G1/S or a basal promoter with six Forkhead DNA-binding sites with
Jeffrey T J Huang et al.
International journal of oncology, 25(3), 751-757 (2004-08-04)
We describe here the identification and characterization of a human forkhead box gene, FOXK1, that encodes predicted proteins most homologous to the mouse myocyte nuclear factor (MNF)/forkhead box K1 (FoxK1). Human FOXK1 is located at the chromosomal position 7p22. Two

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service