Skip to Content
Merck
All Photos(6)

Key Documents

HPA017060

Sigma-Aldrich

Anti-BST2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-BST-2, Anti-Bone marrow stromal antigen 2, Anti-CD317 antigen, Anti-HM1.24 antigen

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

THLLQQELTEAQKGFQDVEAQAATCNHTVMALMASLDAEKAQGQKKVEELEGEITTLNHKLQDASAEVERLRRENQVLSVRIADKKYYPSSQDSSSAAAPQLLIVLLG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BST2(684)

General description

BST2 (bone marrow stromal cell antigen 2) is an interferon-induced cellular protein expressed on a subset of medullary thymocytes. It is composed of an N-terminal cytoplasmic domain, a transmembrane domain, an ectodomain, and a C-terminal membrane anchor. In its N-terminal end, it consists of three cysteine residues, responsible for the formation of cysteine-linked dimers.

Immunogen

Bone marrow stromal antigen 2 Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

BST2 (bone marrow stromal cell antigen 2) plays an important role in the virus-host cell interactions. It functions as a restriction factor viruses to the host cell. It has been reported that BST2 possesses an important clinical role in oral cavity squamous cell carcinoma (OSCC).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70147

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quoc Thang Pham et al.
Anticancer research, 37(6), 2853-2860 (2017-05-30)
Renal cell carcinoma (RCC) is one of the most common types of cancer in developed countries. Bone marrow stromal cell antigen 2 (BST2) gene, which encodes BST2 transmembrane glycoprotein, is overexpressed in several cancer types. In the present study, we
Marta Epeldegui et al.
European journal of immunology, 45(3), 728-737 (2014-11-20)
In contrast to peripheral plasmacytoid DCs (pDCs), thymic pDCs constitutively express low levels of IFN-α. This leads to induction of interferon secondary genes (ISGs) in medullary thymocytes, raising the question whether IFN-α may play a role in T-cell development. When
Sarah Welbourn et al.
The Journal of biological chemistry, 290(6), 3740-3751 (2014-12-20)
BST-2/tetherin is a cellular host factor capable of restricting the release of a variety of enveloped viruses, including HIV-1. Structurally, BST-2 consists of an N-terminal cytoplasmic domain, a transmembrane domain, an ectodomain, and a C-terminal membrane anchor. The BST-2 ectodomain
Sum-Fu Chiang et al.
Disease markers, 2015, 874054-874054 (2015-10-27)
The cancer cell secretome has been recognized as a valuable reservoir for identifying novel serum/plasma biomarkers for different cancers, including colorectal cancer (CRC). This study aimed to verify four CRC cell-secreted proteins (tumor-associated calcium signal transducer 2/trophoblast cell surface antigen
Yoshinori Shigematsu et al.
Oncology letters, 14(1), 999-1004 (2017-07-12)
Bladder cancer, the majority of which is urothelial carcinoma (UC), is a common malignancy worldwide. Genes encoding transmembrane/secretory proteins expressed specifically in certain cancers may be ideal biomarkers for cancer diagnosis and may represent therapeutic targets. In the present study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service