Skip to Content
Merck
All Photos(5)

Key Documents

HPA015801

Sigma-Aldrich

Anti-NSD2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

KMT3G, MMSET, WHSC1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

SANGKTPSCEVNRECSVFLSKAQLSSSLQEGVMQKFNGHDALPFIPADKLKDLTSRVFNGEPGAHDAKLRFESQEMKGIGTPPNTTPIKNGSPEIKLKITKTYMNGKPLFESSICGD

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... WHSC1(7468)

General description

WHSC1 is an oncogene that regulates cell adhesion, cell cycle advancement and malignant transformation in multiple myeloma. This gene also modulates cellular response to DNA damage and inhibits the synthesis of IL-5 mRNA. Loss of WHSC1 has been linked to Wolf-Hirschhorn syndrome . Anti-WHSC1 antibody is specific for WHSC1 in humans.

Immunogen

nuclear receptor binding SET domain protein 2 recombinant protein epitope signature tag (PrEST)

Application

Anti-WHSC1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using protein array and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige. Anti-WHSC1 antibody is suitable for use in indirect immunofluorescence.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73693

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peixin Dong et al.
Oncotarget, 9(33), 23253-23263 (2018-05-26)
Although the oncogene MMSET (also known as NSD2 or WHSC1) has an essential role in malignancies, its impact on human endometrial cancer (EC) metastasis and the molecular mechanism of MMSET regulation are largely unknown. We report that MMSET was markedly
C G Garlisi et al.
American journal of respiratory cell and molecular biology, 24(1), 90-98 (2001-01-12)
Human interleukin (IL)-5 gene transcription is regulated by several transcription factor binding sites, including CLE 0, GATA, and a region from position -123 to -92 known as response element (RE)-II. By expression cloning, a partial protein was identified that bound
Heidi Rye Hudlebusch et al.
British journal of haematology, 130(5), 700-708 (2005-08-24)
The frequently detected t(4;14)(p16.3;q32) translocation in multiple myeloma (MM) results in a dysregulation of two potential oncogenes: multiple myeloma SET domain (MMSET) and fibroblast growth factor receptor 3 (FGFR3). As the expression of FGFR3 is undetectable in 30% of the
Jose L R Brito et al.
Haematologica, 94(1), 78-86 (2008-12-09)
The recurrent immunoglobulin translocation, t(4;14)(p16;q32) occurs in 15% of multiple myeloma patients and is associated with poor prognosis, through an unknown mechanism. The t(4;14) up-regulates fibroblast growth factor receptor 3 (FGFR3) and multiple myeloma SET domain (MMSET) genes. The involvement
Ildiko Hajdu et al.
Proceedings of the National Academy of Sciences of the United States of America, 108(32), 13130-13134 (2011-07-27)
Wolf-Hirschhorn syndrome (WHS) is a malformation syndrome associated with growth retardation, mental retardation, and immunodeficiency resulting from a hemizygous deletion of the short arm of chromosome 4, called the WHS critical region (WHSC). The WHSC1 gene is located in this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service