Skip to Content
Merck
All Photos(7)

Key Documents

HPA015791

Sigma-Aldrich

Anti-SALL4 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Sal-like protein 4, Anti-Zinc finger protein SALL4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200
western blot: 0.04-0.4 μg/mL

immunogen sequence

HALHSSGAGADTLKTLGSHMSQQVSAAVALLSQKAGSQGLSLDALKQAKLPHANIPSATSSLSPGLAPFTLKPDGTRVLPNVMSRLPSALLPQAPGSVLFQSPFSTVALDTSKKGKGKPPNISAVDVKPKDE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SALL4(57167)

Immunogen

Sal-like protein 4 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Sal-like protein 4 is a protein encoded by the SALL4 gene in humans. It mainly functions in developing different cancers and performing vital embryogenic roles. It is found to be associated with gastric cancer, hepatocellular cancer (HCC) and colorectal cancer (CRC). It has some role in All-trans retinoic acid (ATRA)-based regimes in case of leukemia therapy. It may also act as a biomarker for HCC.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73685

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sima Ardalan Khales et al.
Journal of cancer research and clinical oncology, 141(2), 229-235 (2014-08-27)
Colorectal cancer (CRC) is one of the most common causes of cancer-related death worldwide, and there is an urgent need to identify critical diagnostic and prognostic factors for early detection of the disease. Our aim in this study was to
Ta-Chiang Liu et al.
The American journal of surgical pathology, 38(7), 966-972 (2014-05-09)
Prognostic biomarkers that stratify patients with cancer are needed. Recent studies from Asia have implicated SALL4, a stem cell marker, as useful in identifying aggressive cases of hepatocellular carcinoma (HCC), and >50% of the cases tested had upregulation by microarray
Li Liu et al.
The Journal of biological chemistry, 290(17), 10599-10609 (2015-03-05)
All-trans retinoic acid (ATRA) is a differentiation agent that revolutionized the treatment of acute promyelocytic leukemia. However, it has not been useful for other types of acute myeloid leukemia (AML). Here we explored the effect of SALL4, a stem cell
Jie Liu et al.
Hepato-gastroenterology, 61(133), 1459-1464 (2014-12-02)
SALL4 [encoding the Sal-Like 4 (Drosophila) protein, also known as spalt-like transcription factor 4] plays crucial roles in the development of different cancers, as well as vital embryogenic roles. However, knowledge of its relationship to gastric cancer is limited, and
Su-xia Han et al.
Journal of immunology research, 2014, 262385-262385 (2014-05-27)
Sal-like protein 4 (SALL4), is reexpressed in tissues of a subgroup of HCC associated with poor prognosis. Reports of SALL4 serological levels linked to HCC patients are meager and unclear in the prognosis of this malignancy. Immunohistochemistry and optical microscopy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service