Skip to Content
Merck
All Photos(5)

Key Documents

HPA015587

Sigma-Aldrich

Anti-TMEFF2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Hyperplastic polyposis protein 1, Anti-TR-2, Anti-Tomoregulin-2 precursor, Anti-Transmembrane protein with EGF-like and two follistatin-like domains

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50- 1:200

immunogen sequence

FPTSLSDCQTPTGWNCSGYDDRENDLFLCDTNTCKFDGECLRIGDTVTCVCQFKCNNDYVPVCGSNGESYQNECYLRQAACKQQSEILVVSEGSCATDAGSGSGDGVHEGSGETSQKETSTCDICQFGAECDEDA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TMEFF2(23671)

General description

Tomoregulin-2 is a single-pass transmembrane protein encoded by the TMEFF2 gene in humans and is expressed in brain and prostate. It contains an altered epidermal growth factor (EGF)-like motif, two follistatin-like domains, and a cytosolic tail with a putative G-protein-activating motif.

Immunogen

Tomoregulin-2 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Tomoregulin-2 functions in cell signaling, neuronal cell survival, and tumor suppression. It also helps in regulating (i.e. inhibiting) as the non-canonical activin/BMP4 signaling, PI3K, and Ras/ERK1/2 pathways. It contributes to significant inhibition of transcription of Pomc and is used for the treatment of corticotrope tumors. Down-regulation of this gene is observed in human gastric carcinogenesis and negatively regulates STAT3. TMEFF2 may act as a biomarker for carcinogenesis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72667

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joshua M Corbin et al.
The Prostate, 76(1), 97-113 (2015-09-30)
Previous results from our lab indicate a tumor suppressor role for the transmembrane protein with epidermal growth factor and two follistatin motifs 2 (TMEFF2) in prostate cancer (PCa). Here, we further characterize this role and uncover new functions for TMEFF2
Constantin Georgescu et al.
BMC cancer, 19(1), 423-423 (2019-05-08)
The clinical behavior of prostate cancer (PCa) is variable, and while the majority of cases remain indolent, 10% of patients progress to deadly forms of the disease. Current clinical predictors used at the time of diagnosis have limitations to accurately
Tian-Tian Sun et al.
International journal of cancer, 136(5), 1053-1064 (2014-07-06)
The transmembrane protein with epidermal growth factor and two follistatin motifs 2 (TMEFF2) is a single-pass transmembrane protein, and it is downregulated in human gastric cancer and levels correlate with tumor progression and time of survival. However, the mechanism of
Joshua M Corbin et al.
Molecular therapy. Nucleic acids, 24, 337-351 (2021-04-15)
Resistance to anti-androgen therapy in prostate cancer (PCa) is often driven by genetic and epigenetic aberrations in the androgen receptor (AR) and coregulators that maintain androgen signaling activity. We show that specific small RNAs downregulate expression of multiple essential and
Marta Labeur et al.
Journal of molecular endocrinology, 54(1), 51-63 (2015-01-13)
TMEFF2 is a transmembrane protein with unknown function, containing an altered epidermal growth factor (EGF)-like motif, two follistatin-like domains, and a cytosolic tail with a putative G-protein-activating motif. TMEFF2 is predominantly expressed in brain and prostate and has been implicated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service