Skip to Content
Merck
All Photos(2)

Key Documents

HPA014933

Sigma-Aldrich

Anti-TRIM47 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-GOA, Anti-RNF100, Anti-TRIM70, Anti-tripartite motif-containing 47

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

PQEPYDRGRLGRNAHSCCLQWNGRSFSVWFHGLEAPLPHPFSPTVGVCLEYADRALAFYAVRDGKMSLLRRLKASRPRRGGIPASPIDPFQSRLDSHFAGLFTHRLK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TRIM47(91107)

General description

TRIM47 (tripartite motif containing 47) gene is located on human chromosome 17q25. The encoded protein is also called GOA (gene overexpressed in astrocytoma) and is a ring finger B-box coiled-coil (RBCC) protein. Normally, it is highly expressed in kidney and brain, and in other tissues it has a low level of expression. It is also expressed in fetal astrocytes, but not in mature astrocytes. It contains two LXXLL motifs.

Immunogen

tripartite motif-containing 47 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

TRIM47 (tripartite motif containing 47) is up-regulated in astrocytomas, and studies suggest that it is involved in astrocytoma tumorigenesis, by playing a role in astrocytoma dedifferentiation. It might also be related to other cancer types.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72617

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yudong Han et al.
Oncotarget, 8(14), 22730-22740 (2017-02-12)
Non-small cell lung carcinoma (NSCLC) is the most common malignancy with the highest morbidity and mortality. In this study, we found that tripartite motif containing 47 (TRIM47) expression level was higher in tumor tissues than in normal adjacent tissues. Overexpression
D A Vandeputte et al.
Biochemical and biophysical research communications, 286(3), 574-579 (2001-08-21)
Serial analysis of gene expression (SAGE) was used to identify a gene named GOA (gene overexpressed in astrocytoma), which codes for a novel Ring finger B-box coiled-coil (RBCC) protein. Northern blot hybridization showed overexpression of GOA in 9 of 10
Helena Schmidt et al.
Experimental gerontology, 47(11), 873-877 (2012-06-28)
Subcortical vascular dementia or cerebral small vessel disease is a common cause of disability in the elderly. On magnetic resonance imaging the disease is manifested as white matter lesions, lacunes and microbleeds. Its etiology is complex, with age and hypertension

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service