Skip to Content
Merck
All Photos(9)

Key Documents

HPA012604

Sigma-Aldrich

Anti-ALDH18A1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Δ1-pyrroline-5-carboxylate synthetase antibody produced in rabbit, Anti-Aldehyde dehydrogenase 18 family member A1 antibody produced in rabbit, Anti-P5CS antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, rat, human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

SVTFGTKSRVGMGGMEAKVKAALWALQGGTSVVIANGTHPKVSGHVITDIVEGKKVGTFFSEVKPAGPTVEQQGEMARSGGRMLATLEPEQRAEIIHHLADLLTDQRD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ALDH18A1(5832)

Looking for similar products? Visit Product Comparison Guide

General description

Aldehyde dehydrogenase 18 family member A1 (ALDH18A1) is localized to the mitochondrial inner membrane. It is an ATP- and NADPH-dependent bifunctional enzyme. The gene encoding this protein is present on chromosome 10.

Immunogen

Delta-1-pyrroline-5-carboxylate synthetase recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Aldehyde dehydrogenase 18 family member A1 (ALDH18A1) catalyzes the reduction of glutamate to Δ1-pyrroline-5-carboxylate which is a critical step in the de-novo biosynthesis of proline, arginine and ornithine. It is a bifunctional enzyme having both γ-glutamyl kinase (γ-GK) and γ-glutamyl phosphate reductase (γ-GPR) activities. The enzyme undergoes alternative splicing to give rise to two isoforms which differ by a two-amino acid insert at the N-terminus of the γ-glutamyl kinase active site. The short isoform is highly active in the gut and it takes part in arginine biosynthesis. The long isoform is involved in the synthesis of proline from glutamate and is expressed in many tissues.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71373

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Madeleine L Craze et al.
British journal of cancer, 118(2), 258-265 (2017-11-24)
Altered cellular metabolism is a hallmark of cancer and some are reliant on glutamine for sustained proliferation and survival. We hypothesise that the glutamine-proline regulatory axis has a key role in breast cancer (BC) in the highly proliferative classes. Glutaminase
C A Hu et al.
The Journal of biological chemistry, 274(10), 6754-6762 (1999-02-26)
Delta1-Pyrroline-5-carboxylate synthase (P5CS; EC not assigned), a mitochondrial inner membrane, ATP- and NADPH-dependent, bifunctional enzyme, catalyzes the reduction of glutamate to Delta1-pyrroline-5-carboxylate, a critical step in the de novo biosynthesis of proline and ornithine. We utilized published plant P5CS sequence
M R Baumgartner et al.
Human molecular genetics, 9(19), 2853-2858 (2000-11-25)
delta(1)-pyrroline-5-carboxylate synthase (P5CS), a bifunctional ATP- and NADPH-dependent mitochondrial enzyme, catalyzes the reduction of glutamate to delta(1)-pyrroline-5-carboxylate, a critical step in the biosynthesis of proline, ornithine and arginine. Recently, we reported the cloning and expression of human and murine P5CS
James D Budge et al.
Metabolic engineering communications, 13, e00179-e00179 (2021-08-14)
Chinese hamster ovary (CHO) cells are the leading mammalian cell host employed to produce complex secreted recombinant biotherapeutics such as monoclonal antibodies (mAbs). Metabolic selection marker technologies (e.g. glutamine synthetase (GS) or dihydrofolate reductase (DHFR)) are routinely employed to generate
Zhaoying Yang et al.
Cell death and differentiation, 28(1), 303-319 (2020-08-10)
Pyrroline-5-carboxylate synthase (P5CS) catalyzes the synthesis of pyrroline-5-carboxylate (P5C), a key precursor for the synthesis of proline and ornithine. P5CS malfunction leads to multiple human diseases; however, the molecular mechanism underlying these diseases is unknown. We found that P5CS localizes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service