Skip to Content
Merck
All Photos(2)

Key Documents

HPA011906

Sigma-Aldrich

Anti-CD151 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CD151 antigen, Anti-GP27, Anti-Membrane glycoprotein SFA-1, Anti-PETA-3, Anti-Platelet-endothelial tetraspan antigen 3, Anti-Tetraspanin-24, Anti-Tspan-24

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

LNTELKENLKDTMTKRYHQPGHEAVTSAVDQLQQEFHCCGSNNSQDWRDSEWIRSQEAGGRVVPDSCCKTVVALCGQRDHASNIYKVEGGCITKLETF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CD151(977)

Immunogen

CD151 antigen recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CD151 is a transmembrane protein belonging to the tetraspanin family. It is involved in the regulation of cell-substrate adhesion, cell migration, tumor progression, cell metastasis, and angiogenesis. It possesses high binding affinity for the laminin-binding integrins (α3β1, α6β1 and α6β4) and other tetraspanins including growth factors (HGFR, EGFR and TGF-β1R) and matrix metalloproteinases (MMP-7, MMP-2 and MMP-9). It also controls the post-adhesion activities including cell spreading, migration, formation of metastasis, and invasion. In neoplastic and endothelial cells, it participates in tumor neovascularization. Positive expression of CD151 is associated with several types of cancers and research shows the potentiality of the gene as a prognostic biomarker of gallbladder cancer (GBC), gastric cancers.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72489

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hassan Vahidnezhad et al.
Matrix biology : journal of the International Society for Matrix Biology, 66, 22-33 (2017-11-16)
Epidermolysis bullosa (EB) is caused by mutations in as many as 19 distinct genes. We have developed a next-generation sequencing (NGS) panel targeting genes known to be mutated in skin fragility disorders, including tetraspanin CD151 expressed in keratinocytes at the
Sang Yun Ha et al.
Annals of surgical oncology, 21(4), 1099-1106 (2013-12-07)
CD151, a transmembrane protein of the tetraspanin family, is implicated in the regulation of cell-substrate adhesion and cell migration. Overexpression of CD151 has been reported in several cancers and controls MET-dependent neoplastic growth by enhancing receptor signaling. However, association of
Noriko Matsumoto et al.
Surgery, 156(5), 1212-1217 (2014-06-04)
CD151 is a member of the tetraspanin family, which interacts with laminin-binding integrins and other tetraspanins. CD151 is involved in several pathologic activities associated with tumor progression, including metastasis and angiogenesis. The aim of this study was to clarify the
Rafal Sadej et al.
Laboratory investigation; a journal of technical methods and pathology, 94(1), 41-51 (2013-11-20)
Originally identified as a molecular organizer of interacting proteins into tetraspanin-enriched microdomains, the tetraspanin CD151 has now been shown to be involved in tumour progression. Increasing evidence emerging from in vitro, in vivo and clinical analyses implicates this tetraspanin in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service