Skip to Content
Merck
All Photos(7)

Key Documents

HPA011105

Sigma-Aldrich

Anti-SFN antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-14-3-3 protein sigma, Anti-Epithelial cell marker protein 1, Anti-Stratifin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, mouse, rat

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

LSVAYKNVVGGQRAAWRVLSSIEQKSNEEGSEEKGPEVREYREKVETELQGVCDTVLGLLDSHLIKEAGDAESRVFYLKMKGDYYRYLAEVATGDDKKRIIDSARSAYQEAMDISKKEMP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SFN(2810)

General description

Stratifin (SFN) belongs to the 14-3-3 protein family. SFN gene is located on the human chromosome at 1p36.11.

Immunogen

14-3-3 protein sigma recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-SFN antibody produced in rabbit has been used in western blotting.

Biochem/physiol Actions

Stratifin (SFN) protein acts as a biomarker of keratinocyte differentiation. SFN gene functions as a tumor suppressor and inducer of apoptosis. Downregulation of the stratifin gene is associated with breast cancer. SFN gene is considered as a p53-inducible gene that reacts to DNA-damaging agents. SFN acts as a negative regulator of cell cycle progression. Overexpression of the SFN gene leads to lung adenocarcinoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71125

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samatha Bhat et al.
Biochimie, 121, 298-311 (2016-01-09)
Cervical cancer is the second most common cancer among women worldwide. About 528,000 women are diagnosed with cervical cancer contributing to around 266,000 deaths, across the globe every year. Out of these, the burden of 226,000 (85%) deaths occurs in
Ajay P Singh et al.
Cancer letters, 259(1), 28-38 (2007-11-06)
Previously, we have developed a unique in vitro LNCaP cell model, which includes androgen-dependent (LNCaP-C33), androgen-independent (LNCaP-C81) and an intermediate phenotype (LNCaP-C51) cell lines resembling the stages of prostate cancer progression to hormone independence. This model is advantageous in overcoming
Jun Gu et al.
International immunopharmacology, 32, 1-7 (2016-01-18)
Resveratrol (RSV) has many biological effects, including antitumor and antiviral activities, and vascular protection. Recent studies have suggested that RSV exerts its antitumor effects through induction of autophagy by an unknown mechanism. Doxorubicin (DOX) is a wide spectrum antitumor drug
Martin L Biniossek et al.
Journal of proteomics, 91, 515-535 (2013-08-24)
Telomerase inhibition causes progressive telomere shortening and cellular senescence, which constitutes a universal barrier to tumor growth and therefore an attractive target for tumor therapy. To expand our previous studies, we investigated the global effects of telomere dysfunction on the
C B Umbricht et al.
Oncogene, 20(26), 3348-3353 (2001-06-26)
We have identified 14-3-3 sigma (sigma) as a gene whose expression is lost in breast carcinomas, primarily by methylation-mediated silencing. In this report, we investigated the timing of loss of sigma gene expression during breast tumorigenesis in vivo. We analysed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service