Skip to Content
Merck
All Photos(5)

Key Documents

HPA010978

Sigma-Aldrich

Anti-CDCP1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CD318 antigen, Anti-CUB domain-containing protein 1 precursor, Anti-Membrane glycoprotein gp140, Anti-SIMA135, Anti-Subtractive immunization M plus HEp3-associated 135 kDa protein, Anti-Transmembrane and associated with src kinases

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

SKHKISFLCDDLTRLWMNVEKTISCTDHRYCQRKSYSLQVPSDILHLPVELHDFSWKLLVPKDRLSLVLVPAQKLQQHTHEKPCNTSFSYLVASAIPSQDLYFGSFCPGGSIKQIQVKQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDCP1(64866)

General description

CUB-domain-containing protein 1 (CDCP1) is a type I transmembrane protein which functions as a substrate for Src kinases. It has a molecular weight of 140kDa, and has a proteolytically cleaved form of molecular weight 85kDa. It has an intracellular region made of five tyrosines, and a large extracellular region containing the CUB (C1r/C1s, Uegf, Bmp1) domain. Both the isoforms of CDCP1 are expressed in varying ratios in epithelial cells. This gene is localized to human chromosome 3p21.31. CDCP1 protein is composed of 836 amino acids, with a signal peptide of 29 amino acids, and the cytoplasmic domain consists of 150 amino acids.

Immunogen

CUB domain-containing protein 1 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CUB domain-containing protein 1 (CDCP1) is phosphorylated by Src kinases and mediates resistance to anoikis, promotes degradation of extracellular matrix, tumor invasion and metastasis. It is a cancer associated gene and is up-regulated in multiple cancers such as, lung, pancreas and kidney. Its overexpression in these cancers is related to poor prognosis. CDCP1 inhibits integrin-mediated cell adhesion by acting as a substrate of Src-kinases. It also facilitates the metastasis of melanoma, and is associated with poor prognosis of renal cell carcinoma. CDCP1 aids bone morphogenetic protein 4 (BMP4) in initiating epithelial-mesenchymal transition, and suppresses the epithelial phenotype of pancreatic cancer cells. It confers anoikis resistance to lung cancer cells, by participating in protein kinase Cδ (PKCδ) signaling.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72141

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shin Miura et al.
Experimental cell research, 321(2), 209-218 (2014-01-05)
The prognosis of pancreatic cancer is dismal due to the frequent metastasis and invasion to surrounding organs. Numerous molecules are involved in the malignant behavior of pancreatic cancer cells, but the entire process remains unclear. Several reports have suggested that
Wei-Chung Shia et al.
BMC genomics, 12 Suppl 3, S23-S23 (2012-03-06)
Cardiovascular disease is the chief cause of death in Taiwan and many countries, of which myocardial infarction (MI) is the most serious condition. Hyperlipidemia appears to be a significant cause of myocardial infarction, because it causes atherosclerosis directly. In recent
Takamasa Uekita et al.
Cancer science, 102(11), 1943-1948 (2011-08-05)
Tumor metastasis is a complex multistep process by which cells from the primary tumor invade tissues, move through the vasculature, settle at distant sites and eventually grow to form secondary tumors. Altered tyrosine phosphorylation signals in cancer cells contribute to
Mineo Iwata et al.
PloS one, 9(10), e109304-e109304 (2014-10-03)
In vitro expanded bone marrow stromal cells contain at least two populations of fibroblasts, a CD146/MCAM positive population, previously reported to be critical for establishing the stem cell niche and a CD146-negative population that expresses CUB domain-containing protein 1 (CDCP1)/CD318.
Danislav S Spassov et al.
Molecular and cellular biology, 31(4), 766-782 (2010-12-30)
Trask is a recently described transmembrane substrate of Src kinases whose expression and phosphorylation has been correlated with the biology of some cancers. Little is known about the molecular functions of Trask, although its phosphorylation has been associated with cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service