Skip to Content
Merck
All Photos(4)

Key Documents

HPA009174

Sigma-Aldrich

Anti-VAPA antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-33 kDa Vamp-associated protein antibody produced in rabbit, Anti-VAMP-A antibody produced in rabbit, Anti-VAMP-associated protein A antibody produced in rabbit, Anti-VAP-33 antibody produced in rabbit, Anti-VAP-A antibody produced in rabbit, Anti-Vesicle-associated membrane protein-associated protein A antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

RNAi knockdown
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

NDKLGITPPGNAPTVTSMSSINNTVATPASYHTKDDPRGLSVLKQEKQKNDMEPSKAVPLNASKQDGPMPKPHSVSLNDTETRKLMEECKRLQGEMMKLSEENRHLRDEGLRLRKVAHSDKPGSTSTASF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VAPA(9218)

General description

VAPA (vesicle associated membrane protein associated protein A) gene is localized to human chromosome 18p11. The encoded protein is also called VAP33, which shows a wide range of tissue expression. It is an integral membrane protein, which is composed of coiled-coil domains. This protein was originally isolated from Aplysia as an interacting partner of VAMP/synaptobrevin. This protein co-localizes with occluding at the tight junctions in cells. This protein has a molecular weight of 33kDa.

Immunogen

Vesicle-associated membrane protein-associated protein A recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

VAPA (vesicle associated membrane protein associated protein A) plays an essential role in vesicular neurotransmission, where it interacts with presynaptic proteins. Studies suggest that SNPs (single nucleotide polymorphisms) in the 3′UTR (untranslated region) of this gene are linked with bipolar disorder. It is essential for the replication of HCV (hepatitis C virus), where it interacts with HCV nonstructural protein 5A (NS5A) in a G protein pathway suppressor 2 (GPS2)-mediated manner. As this protein binds with VAMP/synaptobrevin, it is thought to be involved in vesicle docking and fusion.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71717

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Styliani Panagiotou et al.
Cell reports, 43(4), 113992-113992 (2024-03-27)
Insulin is packaged into secretory granules that depart the Golgi and undergo a maturation process that involves changes in the protein and lipid composition of the granules. Here, we show that insulin secretory granules form physical contacts with the endoplasmic
Norito Tamura et al.
The Journal of biological chemistry, 297(5), 101338-101338 (2021-10-25)
The lipid molecule ceramide is transported from the endoplasmic reticulum to the Golgi apparatus for sphingomyelin production via the ceramide transport protein (CERT), encoded by CERT1. Hyperphosphorylation of CERT's serine-repeat motif (SRM) decreases its functionality. Some forms of inherited intellectual
Guodong Xu et al.
PloS one, 8(11), e78195-e78195 (2013-11-14)
Hepatitis C virus (HCV) nonstructural protein 5A (NS5A) is a component of the replication complex associated with various cellular proteins. It has been reported that G protein pathway suppressor 2 (GPS2) is a potential NS5A-binding factor, as identified in a
Falk W Lohoff et al.
Journal of neural transmission (Vienna, Austria : 1996), 115(9), 1339-1345 (2008-07-31)
Linkage studies in bipolar disorder (BPD) suggest that a susceptibility locus exists on chromosome 18p11. The vesicle-associated membrane protein-associated protein A (VAPA) gene maps to this region. VAPA interacts with presynaptic proteins and is necessary for vesicular neurotransmission. Dysregulation of
Nicholas L Cianciola et al.
Journal of virology, 91(6) (2017-01-13)
Human adenoviruses (Ads) generally cause mild self-limiting infections but can lead to serious disease and even be fatal in high-risk individuals, underscoring the importance of understanding how the virus counteracts host defense mechanisms. This study had two goals. First, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service