Skip to Content
Merck
All Photos(6)

Key Documents

HPA008848

Sigma-Aldrich

Anti-MCAM antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CD146 antigen antibody produced in rabbit, Anti-Cell surface glycoprotein MUC18 precursor antibody produced in rabbit, Anti-Cell surface glycoprotein P1H12 antibody produced in rabbit, Anti-Melanoma cell adhesion molecule antibody produced in rabbit, Anti-Melanoma-associated antigen A32 antibody produced in rabbit, Anti-Melanoma-associated antigen MUC18 antibody produced in rabbit, Anti-S-endo 1 endothelial-associated antigen antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

LNYRLPSGNHMKESREVTVPVFYPTEKVWLEVEPVGMLKEGDRVEIRCLADGNPPPHFSISKQNPSTREAEEETTNDNGVLVLEPARKEHSGRYECQGLDLDTMISLLSEPQELLVNYVSDVRVSPAAPERQEGSSLTLTCEAESSQDLE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MCAM(4162)

General description

MCAM (melanoma cell adhesion molecule) is a vascular endothelium expressed cell adhesion molecule. It is ectopically expressed by malignant melanoma cells. It belongs to the immunoglobulin (Ig) superfamily. MCAM gene is a single copy gene which is localized to human chromosome 11q23, spanning 15kb and containing 16 exons. The encoded product is a transmembrane glycoprotein with a molecular weight of 113-119kDa. It is also expressed by smooth muscle cells and a subtype of activated T-cells.

Immunogen

Cell surface glycoprotein MUC18 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

MCAM (melanoma cell adhesion molecule) gene is linked to higher relapse probabilities in melanoma patients. It is thought to play roles in signal transduction, development, cell migration, differentiation of mesenchymal stem cells, angiogenesis and inflammatory responses. In mice, this gene is linked with tumor thickness and metastatic capabilities of human melanoma cells. The expression of this gene has potential as a prognostic marker, where the expression could correlate to poor or good outcome in melanoma patients. In late-stage osteoarthritis, this protein has potential as a marker for cartilage progenitor cell population. In gastric cancer (GC) patients, the expression of this gene is linked with lymph node metastasis and poor prognosis. Its expression is also linked to the loss of E-cadherin, and epithelial marker, and the gain of mesenchymal markers nuclear β-catenin and vimentin. Thus, in GC, this gene might be linked with epithelial-mesenchymal transition (EMT) and tumor progression.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71658

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Henry J M Ferguson et al.
Oncotarget, 7(15), 20440-20454 (2016-03-05)
Current vascular-targeted therapies in colorectal cancer (CRC) have shown limited benefit. The lack of novel, specific treatment in CRC has been hampered by a dearth of specific endothelial markers. Microarray comparison of endothelial gene expression in patient-matched CRC and normal
Wen-Fang Liu et al.
International journal of molecular sciences, 13(5), 6399-6406 (2012-07-04)
CD146 has been regarded as a novel potential therapeutic target for multiple cancers. The aim of the study was to investigate the expression of CD146 in gastric cancer and evaluate its clinical-pathological and prognostic significance. The expression of CD146 and
Maria Cristina Rapanotti et al.
Archives of dermatological research, 306(6), 527-537 (2014-06-07)
MCAM/MUC18 is a cell adhesion molecule associated with higher incidence of relapse in melanoma. The purpose of our study was to evaluate its role as a promising disease biomarker of progression through sequential molecular MCAM/MUC18 RT-PCR assay on serial blood
Christopher J Hanley et al.
Nature communications, 14(1), 387-387 (2023-02-01)
Fibroblasts are poorly characterised cells that variably impact tumour progression. Here, we use single cell RNA-sequencing, multiplexed immunohistochemistry and digital cytometry (CIBERSORTx) to identify and characterise three major fibroblast subpopulations in human non-small cell lung cancer: adventitial, alveolar and myofibroblasts.
Joseph W Wragg et al.
Cancer research, 76(8), 2314-2326 (2016-02-28)
The structure and molecular signature of tumor-associated vasculature are distinct from those of the host tissue, offering an opportunity to selectively target the tumor blood vessels. To identify tumor-specific endothelial markers, we performed a microarray on tumor-associated and nonmalignant endothelium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service