Skip to Content
Merck
All Photos(7)

Key Documents

HPA007724

Sigma-Aldrich

Anti-PROS1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Vitamin K-dependent protein S precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

QGASGIKEIIQEKQNKHCLVTVEKGSYYPGSGIAQFHIDYNNVSSAEGWHVNVTLNIRPSTGTGVMLALVSGNNTVPFAVSLVDSTSEKSQDILLSVENTVIYRIQALSLCSDQQSHLEFRVNR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PROS1(5627)

General description

PROS1 (protein S alpha) gene is the active PS (Protein S) gene localized to human chromosome 3p 11.1 - 3q 1 1.2. Another gene PS β (PROS2) is a pseudogene. Exon I of this gene encodes the signal peptide, exon II encodes the propeptide and Gla domain, and exon III encodes a domain with a high aromatic amino acid content. The protein contains a thrombin-sensitive loop encoded by exon IV, four epidermal growth factor-like domains by exons V-VIII, and a sex hormone binding globulin-homologous domain encoded by the last seven exons.

Immunogen

Vitamin K-dependent protein S precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-PROS1 antibody produced in rabbit has been used for global protein profiling to find new molecular biomarkers for common, multifactorial disorders.
Anti-PROS1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

PROS1 (protein S alpha) gene encodes a vitamin K-dependent plasma glycoprotein that functions as a cofactor to activated protein C (APC) in the degradation of coagulation factors Va and VIIIa to inhibit blood coagulation in the presence of phospholipids and Ca2+ ions. The protein is active when it is circulating in the plasma in a free form. When bound to C4b-binding protein (C4b-BP), it circulates in the plasma in an inactive form. Only the active form serves as a cofactor to APC. Mutations in this gene may cause free protein S (type IIa) deficiency resulting in autosomal dominant hereditary thrombophilia.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST84444

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Duchemin et al.
Blood, 86(9), 3436-3443 (1995-11-01)
A Ser 460 to Pro mutation of protein S (PS), involving a T to C transition in exon XIII of the protein S alpha (PROS1) gene and known as the Heerlen polymorphism, was found in 16 of 85 symptomatic patients
Bernet S Kato et al.
Proteome science, 9, 73-73 (2011-11-19)
The advent of affinity-based proteomics technologies for global protein profiling provides the prospect of finding new molecular biomarkers for common, multifactorial disorders. The molecular phenotypes obtained from studies on such platforms are driven by multiple sources, including genetic, environmental, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service