Skip to Content
Merck
All Photos(7)

Key Documents

HPA006811

Sigma-Aldrich

Anti-ZBTB7B antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-T-helper-inducing POZ/Krueppel-like factor, Anti-Zfp-67, Anti-Zinc finger and BTB domain-containing protein 15, Anti-Zinc finger and BTB domain-containing protein 7B, Anti-Zinc finger protein 67 homolog, Anti-Zinc finger protein Th-POK, Anti-hcKrox

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43
conjugate:
unconjugated
application:
IF
IHC
clone:
polyclonal
species reactivity:
human
citations:
4
technique(s):
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

AHPLTYEEEEVAGRVGSSGGSGPGDSYSPPTGTASPPEGPQSYEPYEGEEEEEELVYPPAYGLAQGGGPPLSPEELGSDEDAIDPDLMAYLSSLHQDNLAPGLDSQDKLVRKRRSQMPQECPVCHKII

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZBTB7B(51043)

General description

ZBTB7B (zinc finger and BTB domain containing 7B) is a zinc finger-containing transcription factor belonging to the early growth response gene family. It is expressed in the foreskin and dermal fibroblasts. It consists of three kruppel-like zinc-finger DNA binding domains, a POZ (poxvirus and zinc finger) domain for the transcriptional repression and protein dimerization.

Immunogen

Zinc finger and BTB domain-containing protein 7B recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ZBTB7B (zinc finger and BTB domain containing 7B) is highly essential for the CD4+ commitment and prevention of CD8+ commitment. ZBTB7B plays an important role in the development of T cell subsets such as double-negative invariant natural killer T (iNKT) cells, γδ cells, and memory CD8+ T cells. It also represses collagen transcription by binding to the α1 and α2 collagen gene promoters directly. It has been reported that ZBTB7B may play a role in the repression of fibronectin and other matrix genes.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70952

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francesco Mariani et al.
PloS one, 8(1), e54488-e54488 (2013-01-26)
We purposed to evaluate the role of Th inducing POZ-Kruppel Factor (ThPOK), a transcriptional regulator of T cell fate, in tumour-induced immune system plasticity in colorectal carcinogenesis. The amounts of CD4+, CD8+ and CD56+ and ThPOK+ cells infiltrate in normal
R L Widom et al.
Matrix biology : journal of the International Society for Matrix Biology, 20(7), 451-462 (2001-11-03)
The transcription factor cKrox was originally identified as a protein that bound to a negative transcription regulatory element in the murine alpha1(I) collagen promoter. We recently reported the cloning and characterization of human cKrox (hcKrox). Overexpression of hcKrox in NIH3T3
Dietmar J Kappes
Immunological reviews, 238(1), 182-194 (2010-10-26)
The role of the zinc finger transcription factor ThPOK (T-helper-inducing POZ-Kruppel-like factor) in promoting commitment of αβ T cells to the CD4 lineage is now well established. New results indicate that ThPOK is also important for the development and/or acquisition
R L Widom et al.
Gene, 198(1-2), 407-420 (1997-11-25)
cKrox is a novel zinc finger-containing transcription factor that binds to the alpha1(I) and alpha2(I) collagen gene promoters. The gene coding for cKrox is a new member of a family of early growth response genes, that play important roles in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service