Skip to Content
Merck
All Photos(6)

Documents

HPA006461

Sigma-Aldrich

Anti-FASN antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Fatty acid synthase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

RWLRVTVAGGVHISGLHTESAPRRQQEQQVPILEKFCFTPHTEEGCLSERAALQEELQLCKGLVQALQTKVTQQGLKMVVPGLDGAQIPRDPSQQELPRLLSAACRLQLNGN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FASN(2194)

General description

FASN (fatty acid synthase) is a large, multidomain, lipogenic protein. It is composed of seven catalytic domains, which include β-ketoacyl synthase (KS), malonyl acetyl transferase, dehydratase (DH), enoyl-acyl carrier protein-reductase (ER), β-ketoacyl reductase (KR), acyl carrier protein (ACP), and thioesterase domains. This protein exists in a functionally homodimeric form. Under normal conditions, it is expressed at low levels.

Immunogen

Fatty acid synthase recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

FASN (fatty acid synthase) is responsible for the biogenesis of long chain saturated fatty acids, which make up lipid signaling molecules, membranes and anchors for membrane proteins. This protein is linked to multiple disorders such as, hepatic steatosis, cancer, inflammation, obesity and diabetes. It is highly up-regulated in multiple cancers such as, breast, lung, prostate, colon, pancreatic and bladder. In breast cancer, this protein controls liver fatty acid-binding protein (L-FABP), vascular endothelial growth factor (VEGF) and VEGF Receptor-2 (VEGFR2), thus, promoting epithelial-mesenchymal transition of breast cancer cells. Trastuzumab-resistance in breast cancer is mediated by the up-regulation of FASN by Pin-1 (peptidylprolyl cis/trans isomerase, NIMA-interacting 1) protein. It is up-regulated in malignant gliomas, and might have potential as a therapeutic target for the drug Orlistat.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70489

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katherine H Sippel et al.
The Journal of biological chemistry, 289(48), 33287-33295 (2014-10-11)
Human fatty acid synthase (FAS) is a large, multidomain protein that synthesizes long chain fatty acids. Because these fatty acids are primarily provided by diet, FAS is normally expressed at low levels; however, it is highly up-regulated in many cancers.
Anup S Ramdhave et al.
American journal of translational research, 9(3), 830-844 (2017-04-08)
Emerging evidence suggests that, dysregulation of fatty acid synthase (FASN) and insulin-like growth factor-1 (IGF-1) could play a vital role in pathology of various diseases. Our aim was to determine the changes in FASN and IGF-1 levels concomitant to long
Elisabetta Vergani et al.
Frontiers in cell and developmental biology, 10, 927118-927118 (2022-08-02)
Drug resistance limits the achievement of persistent cures for the treatment of melanoma, in spite of the efficacy of the available drugs. The aim of the present study was to explore the involvement of lipid metabolism in melanoma resistance and
Hyo Jeong Yun et al.
Anticancer research, 34(3), 1409-1416 (2014-03-07)
Clinical trials have shown efficacy of the anti-HER2 monoclonal antibody trastuzumab in metastatic breast cancer patients. The aim of the present study was to elucidate the mechanisms by which up-regulation of fatty acid synthase (FAS) expression confers resistance to trastuzumab
Junqin Li et al.
International journal of biological sciences, 10(2), 171-180 (2014-02-13)
This study aimed to investigate the role of fatty acid synthase (FASN) in the epithelial-mesenchymal transition (EMT) of breast cancer cells. MCF-7 cells and MCF-7 cells overexpressing mitogen-activated protein kinase 5 (MCF-7-MEK5) were used in this study. MCF-7-MEK5 cells showed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service