Skip to Content
Merck
All Photos(3)

Key Documents

HPA005534

Sigma-Aldrich

Anti-GLI3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Zinc finger protein GLI3 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

GQQMLGQISATSHINIYQGPESCLPGAHGMGSQPSSLAVVRGYQPCASFGGSRRQAMPRDSLALQSGQLSDTSQTCRVNGIKMEMKGQPHPLCSNLQNYSGQFYDQTVGFSQQDTKAGSFSISDASCLLQGTSAKNSELLSPGA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GLI3(2737)

General description

GLI family zinc finger 3 (GLI3) belongs to Gli family of DNA binding proteins. There are three Gli proteins (GLI1, GLI2 and GLI3), which contain a DNA-binding domain made up of five zinc fingers. Fingers 4 and 5 recognize the Gli-binding DNA sequence, and fingers 3, 4 and 5 interact closely with DNA. Their C-terminus consists of transcriptional activator domain (TAD), and in GLI3, the repressor domain (RD) is present at the N-terminus. In humans, GLI3 gene is present on chromosome 7p13, and spans 240kb. The encoded protein consists of 1596 amino acids. This protein is expressed during early stages of development.

Immunogen

Zinc finger protein GLI3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

GLI family zinc finger 3 (GLI3) is involved in the sonic hedgehog (SHH) signaling in a downstream manner. It binds to the promoter region of genes downstream to it, and regulates their expression. In forebrain, this protein suppresses ventral cell types and regulates the specification of dorsal cell types. It suppresses digit formation when expressed in the anterior portion of the limb buds. Mutations in GLI3 gene usually lead to loss of function, and are a cause of multiple disorders, such as Greig Cephalopolysyndactyly Syndrome (GCPS), Acrocallosal Syndrome, Pallister-Hall Syndrome, Pre-axial Polydactyly type IV and Post-axial Polydactyly type A. Mutation in the N-terminal of GLI3 gene is linked to esophageal atresia. GLI3 is expressed in mesenchymal and epithelial layers during tooth development. SNP rs929387 of GLI3 is associated with tooth agenesis in Chinese Han population.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST77454

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert Tsanev et al.
PloS one, 8(10), e76972-e76972 (2013-10-23)
The transcription factor Gli3 is acting mainly as a transcriptional repressor in the Sonic hedgehog signal transduction pathway. Gli3 contains a repressor domain in its N-terminus from residue G106 to E236. In this study we have characterized the intracellular structure
A Wild et al.
Human molecular genetics, 6(11), 1979-1984 (1997-09-25)
Greig cephalopolysyndactyly syndrome (GCPS, MIM 175700) is a rare autosomal dominant developmental disorder characterized by craniofacial abnormalities and post-axial and pre-axial polydactyly as well as syndactyly of hands and feet. Human GLI3, located on chromosome 7p13, is a candidate gene
Haochen Liu et al.
PloS one, 8(11), e80860-e80860 (2013-11-28)
Tooth agenesis is one of the most common anomalies of human dentition. Recent studies suggest that a number of genes are related to both syndromic and non-syndromic forms of hypodontia. In a previous study, we observed that polymorphism in rs929387
Michael Volodarsky et al.
BMC medical genetics, 15, 110-110 (2014-10-01)
Polydactyly is a highly common congenital limb defect. Extra digits may appear as an isolated anomaly or as a part of a syndrome. Mutations in GLI3 have been shown to cause Greig cephalopolysyndactyly, Pallister-Hall syndrome and non-syndromic polydactyly. Genotype-phenotype correlation
Lin Yang et al.
Biochimica et biophysica acta, 1842(9), 1755-1761 (2014-05-14)
Esophageal atresia is a common and life-threatening birth defect with a poorly understood etiology. In this study, we analyzed the sequence variants of coding regions for a set of esophageal atresia-related genes including MYCN, SOX2, CHD7, GLI3, FGFR2 and PTEN

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service