Skip to Content
Merck
All Photos(1)

Key Documents

HPA005131

Sigma-Aldrich

Anti-REN antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Angiotensinogenase antibody produced in rabbit, Anti-Renin precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

FHYINLIKTGVWQIQMKGVSVGSSTLLCEDGCLALVDTGASYISGSTSSIEKLMEALGAKKRLFDYVVKCNEGPTLPDISFHLGGKEYTLTSADYVFQESYSSKKLCTLAIHAMDIPPPTGPTWALGATFIRKFYTEFD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... REN(5972)

General description

REN gene encodes the enzyme renin, which is expressed mainly by the granular cells of the kidney in the juxtaglomerular apparatus. REN gene is 12.5kb in length and is localised to the chromosomal region 1q32. Renin is a protease enzyme, which cleaves at the aspartate residue and is synthesised as prorenin. Prorenin includes an additional 43 amino acid fragment at the N-terminal. Renin has a molecular weight of 37kDa and is composed of 340 amino acids. There is evidence that renin is also expressed in ovary, testis, submandibular gland, adrenal gland, brain, heart etc.

Immunogen

Renin precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Renin is a key enzyme in the renin-angiotensin system (RAS), which is responsible for maintaining blood pressure and salt homeostasis. Renin converts the inactive angiotensinogen to angiotensin I and this step is the rate limiting step in RAS. Angiotensin I is then converted into angiotensin II, which, through a series of steps, leads to an increase in blood pressure and sodium retention by kidneys. Mutations, which lead to the deletion of leucine or exchange of leucine in the signal peptide of prorenin, are found to be associated with early onset anemia, hypouricosuric hyperuricemia and progressive kidney failure. Renin rs6693954 polymorphism has been linked with blood pressure in type II diabetes patients, in a gender-dependent manner.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST85921

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

U Mohana Vamsi et al.
Journal of the renin-angiotensin-aldosterone system : JRAAS, 14(3), 242-247 (2012-10-31)
Renin is a rate-limiting enzyme of the renin-angiotensin-aldosterone system (RAAS) that plays a crucial role in the regulation of blood pressure. The renin gene has been suggested as a marker for genetic predisposition to essential hypertension (EHT) in humans. The
Min-Hua Tseng et al.
Cells, 10(4) (2021-05-01)
We has identified a founder homozygous E3_E4 del: 2870 bp deletion + 9 bp insertion in AGT gene encoding angiotensinogen responsible for autosomal recessive renal tubular dysgenesis (ARRTD) with nearly-fatal outcome. High-dose hydrocortisone therapy successfully rescued one patient with an
Liza U Ljungberg et al.
Journal of the renin-angiotensin-aldosterone system : JRAAS, 15(1), 61-68 (2013-01-30)
Patients with type 2 diabetes (T2D) are at high risk of developing hypertension and related cardiovascular disease. The renin-angiotensin system (RAS) plays a central role in regulation of blood pressure (BP). Accordingly, each component of this system represents a potential
Masayoshi Kukida et al.
Arteriosclerosis, thrombosis, and vascular biology, 41(11), 2851-2853 (2021-09-10)
[Figure: see text].
Genevieve Nguyen et al.
The Journal of clinical investigation, 109(11), 1417-1427 (2002-06-05)
Renin is an aspartyl protease essential for the control of blood pressure and was long suspected to have cellular receptors. We report the expression cloning of the human renin receptor complementary DNA encoding a 350-amino acid protein with a single

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service