Skip to Content
Merck
All Photos(4)

Key Documents

HPA004734

Sigma-Aldrich

Anti-MEF2B antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Myocyte enhancer factor 2B, Anti-RSRFR2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

VLLKYTEYSEPHESRTNTDILETLKRRGIGLDGPELEPDEGPEEPGEKFRRLAGEGGDPALPRPRLYPAAPAMPSPDVVYGALPPPGCDPSGLGEALPAQSRPSPFRPAAPKAGPPGLVHPLFSPSHLTSKTPPPLYLPTE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

MEF2B, a member of MADS family, is expressed during early embryonic development. It is also expressed in brain during neuronal survival and differentiation. The member of this family exists in three different states such as helix, sheet and coil with MADS domain.

Immunogen

myocyte enhancer factor 2B recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

MEF2B (Myocyte enhancer factor 2B) is involved in the myogenic transformation of the epithelial cells. In presence of calcium, MEF2B regulates several gene expression in collaboration with Class II histone deacetylases (HDACs). It also regulates a series of gene expression associated with the development of skeletal, cardiac, and smooth muscle.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86897

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiang Sun et al.
Experimental biology and medicine (Maywood, N.J.), 237(2), 178-185 (2012-02-04)
It has recently emerged that the myogenic contribution of the epithelial mesenchymal transition plays a role in neoplastic invasion and metastasis. Myocyte enhancing factor 2B (MEF2B) is the only MEF2 isoform expressed during early embryonic development, and is herein proposed
Aidong Han et al.
Journal of molecular biology, 345(1), 91-102 (2004-11-30)
Class II histone deacetylases (HDACs) bind myocyte enhancer factor-2 (MEF2) and repress specific gene expression in a calcium-dependent manner. Despite their significant physiological functions in muscle, immune and neuronal cells, the mechanism of recruitment of class II HDACs by MEF2
Xin-Jun Guo
Yi chuan = Hereditas, 33(9), 975-981 (2011-09-29)
As one of the members of MADS family, MEF2 group is important in regulating development. Analytical tools of NCBI, ExPASy, CBS, CDD, and SABLE were adopted to analyze the properties of human MEF2 proteins, and evolutionary tree was built according
M Li et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 21(17), 6544-6552 (2001-08-23)
Myocyte enhancer factor 2 (MEF2) proteins are important regulators of gene expression during the development of skeletal, cardiac, and smooth muscle. MEF2 proteins are also present in brain and recently have been implicated in neuronal survival and differentiation. In this
Siraj M El Jamal et al.
Laboratory investigation; a journal of technical methods and pathology, 99(4), 539-550 (2018-11-18)
Myocyte enhancer-binding factor 2B (MEF2B) has been implicated as a transcriptional regulator for BCL6. However, details about the interaction between MEF2B and BCL6 during expression, as well as the relationship of MEF2B to the expression of other germinal center (GC)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service