Skip to Content
Merck
All Photos(4)

Key Documents

HPA004102

Sigma-Aldrich

Anti-TNFRSF1A antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CD120a antigen, Anti-TNF-R1, Anti-TNF-RI, Anti-TNFR-I, Anti-Tumor necrosis factor receptor superfamily member 1A precursor, Anti-p55, Anti-p60

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

GDREKRDSVCPQGKYIHPQNNSICCTKCHKGTYLYNDCPGPGQDTDCRECESGSFTASENHLRHCLSCSKCRKEMGQVEISSCTVDRDTVCGCRKNQYRHYWSENLFQCFNCSLCLNGTVHLSCQEKQNTVCTCHAGFFLRENECVSC

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TNFRSF1A(7132)

Immunogen

Tumor necrosis factor receptor superfamily member 1A precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Tumor necrosis factor receptor superfamily member 1A is a protein encoded by the TNFRSF1A gene in humans. It is mapped to chromosome 12p13. The gene encodes for human tumor necrosis factor receptor. It is a 415 amino acid polypeptide with a single membrane-spanning region. It consist of extracellular cysteine-rich domain that is homologous to the nerve growth factor receptor and the B cell activation protein Bp50. Mutation in this gene cause the autosomal dominant TNF receptor-associated periodic syndrome (TRAPS) which is a systemic auto-inflammatory disorder.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86727

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ola H Negm et al.
European journal of immunology, 44(7), 2096-2110 (2014-03-29)
Mutations in TNFRSF1A encoding TNF receptor 1 (TNFR1) cause the autosomal dominant TNF receptor-associated periodic syndrome (TRAPS): a systemic autoinflammatory disorder. Misfolding, intracellular aggregation, and ligand-independent signaling by mutant TNFR1 are central to disease pathophysiology. Our aim was to understand
Luca Cantarini et al.
Seminars in arthritis and rheumatism, 43(6), 818-823 (2014-01-08)
To analyze the clinical manifestations and response to treatment in a cohort of adult patients presenting with recurrent inflammatory attacks and carrying low-penetrance TNFRSF1A variants, as well as to provide data on their long-term follow-up. We performed a retrospective chart
E Baker et al.
Cytogenetics and cell genetics, 57(2-3), 117-118 (1991-01-01)
TNFR1 and TNFR2, the genes encoding the two forms of the human tumor necrosis factor receptor, were localized to normal human chromosomes by in situ hybridization and Southern blot analysis of a series of human x mouse hybrid cell lines.
T J Schall et al.
Cell, 61(2), 361-370 (1990-04-20)
A human tumor necrosis factor (TNF) binding protein from serum of cancer patients was purified to homogeneity and partially sequenced. Synthetic DNA probes based on amino acid sequence information were used to isolate cDNA clones encoding a receptor for TNF.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service