Skip to Content
Merck
All Photos(7)

Key Documents

HPA003575

Sigma-Aldrich

Anti-TRIM22 antibody produced in rabbit

enhanced validation

Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-50 kDa-stimulated trans-acting factor antibody produced in rabbit, Anti-RING finger protein 94 antibody produced in rabbit, Anti-Staf-50 antibody produced in rabbit, Anti-Tripartite motif-containing protein 22 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

RNAi knockdown
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

NEVVKECQEKLQVALQRLIKEDQEAEKLEDDIRQERTAWKNYIQIERQKILKGFNEMRVILDNEEQRELQKLEEGEVNVLDNLAAATDQLVQQRQDASTLISDLQRRLRGSSVEMLQDVIDVMKRSESWTLKKPKSVSKKLKSVFRV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TRIM22(10346)

Looking for similar products? Visit Product Comparison Guide

Immunogen

Tripartite motif-containing protein 22 recombinant protein epitope signature tag (PrEST)

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

Tripartite motif-containing 22 is a protein encoded by the TRIM22 gene in humans and belongs to tripartite motif (TRIM) protein superfamily. The members of this family are important effectors of the innate immune response against viral infections. It is an interferon (IFN)-inducible protein as well as a p53 target gene. It plays a role in viral defense by restricting viral replication and may function as a ubiquitin E3 ligase. This gene is found to exert antiviral activity against RNA viruses, such as hepatitis B virus (HBV), encephalomyocarditis virus (ECMV) and human immunodeficiency virus type 1 (HIV-1). It is involved in the activation of NF-κB. It acts as a positive regulator of NF-κB-mediated transcription.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86655

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ravesh Singh et al.
Journal of virology, 88(8), 4291-4303 (2014-01-31)
The antiviral role of TRIM E3 ligases in vivo is not fully understood. To test the hypothesis that TRIM5α and TRIM22 have differential transcriptional regulation and distinct anti-HIV roles according to infection phase and compartment, we measured TRIM5α, TRIM22, and
Hui Tian et al.
Yonsei medical journal, 59(4), 511-518 (2018-05-12)
Hepatitis C virus (HCV) infection is a major cause of liver disease. Several miRNAs have been found to be associated with HCV infection. This study aimed to investigate the functional roles and possible molecular mechanisms of miR-215 in HCV replication.
Shanshan Yu et al.
Biochemical and biophysical research communications, 410(2), 247-251 (2011-06-10)
Increasing evidence suggests that TRIM family proteins may play important roles in the regulation of innate immune signaling pathways. Here we report TRIM22 is involved in the activation of NF-κB. It was found that overexpression of TRIM22 could dose-dependently activate
Jessica Petersson et al.
Experimental cell research, 316(4), 568-579 (2009-12-17)
TRIM22 (Staf50), a member of the TRIM protein family, is an interferon (IFN)-inducible protein as well as a p53 target gene. The function of TRIM22 is largely unknown, but TRIM22 is suggested to play a role in viral defense by
Filippo Turrini et al.
Virus research, 269, 197631-197631 (2019-05-29)
The human immunodeficiency virus type-1 (HIV-1) establishes a state of latent infection in a small number of CD4+ T lymphocytes that, nonetheless, represent a major obstacle to viral eradication. We here show that Tripartite Motif-containing protein 22 (TRIM22), an epigenetic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service