Skip to Content
Merck
All Photos(6)

Key Documents

HPA003517

Sigma-Aldrich

Anti-FCGBP antibody produced in rabbit

enhanced validation

Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Fcγ-binding protein antigen antibody produced in rabbit, Anti-FcγBP antibody produced in rabbit, Anti-IgGFc-binding protein precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

GHRFDFQGTCEYLLSAPCHGPPLGAENFTVTVANEHRGSQAVSYTRSVTLQIYNHSLTLSARWPRKLQVDGVFVALPFQLDSLLHAHLSGADVVVTTTSGLSLAFDGDSF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FCGBP(8857)

Looking for similar products? Visit Product Comparison Guide

Immunogen

IgGFc-binding protein precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-FCGBP antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)

Biochem/physiol Actions

IgGFc-binding protein is a protein encoded by the FCGBP gene in humans. The gene is also known as FcγBP. Its mRNA is expressed only in placenta and colonic epithelial cells. It may play an important role in immune protection and inflammation in the intestines of primates. The gene is expressed in both human and mouse prostates. Differential expression of this gene could reflect its potential role in prostate malignancy as well as an indicator for progression of the cancer. It is differentially expressed in normal thyroid tissue, thyroid adenomas and thyroid carcinomas. The gene is constitutively expressed in normal thyroid tissue, its expression is significantly increased in follicular thyroid adenomas and considerably decreased in papillary and follicular thyroid carcinomas. The expression level of this gene in thyroid biopsies is useful for distinguishing between a thyroid follicular adenoma and a follicular carcinoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86466

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nancy A Erickson et al.
PloS one, 10(7), e0131991-e0131991 (2015-07-15)
The secreted, goblet cell-derived protein Clca1 (chloride channel regulator, calcium-activated-1) has been linked to diseases with mucus overproduction, including asthma and cystic fibrosis. In the intestine Clca1 is found in the mucus with an abundance and expression pattern similar to
N Harada et al.
The Journal of biological chemistry, 272(24), 15232-15241 (1997-06-13)
Cloning a cDNA for human IgGFc binding protein (FcgammaBP) from human colonic epithelial cells reveals an mRNA and coding region of 17 and 16.2 kilobases, respectively. The predicted amino acid sequence contains 12 occurrences of a 400-amino acid cysteine-rich unit
Mozammel H Gazi et al.
Cancer biology & therapy, 7(1), 70-75 (2007-10-17)
By means of protein expression profile, mass spectral and/or RT-PCR analyses we found for the first time IgG Fc binding protein (Fc gammaBP), distinct from Fc gamma receptors is expressed in both human and mouse prostates. There is a strong
N O'Donovan et al.
The Journal of endocrinology, 174(3), 517-524 (2002-09-05)
The genetic events involved in thyroid carcinogenesis are still incompletely understood. Several rearrangements and mutations of oncogenes have been implicated in the development of thyroid papillary carcinomas, follicular adenomas and carcinomas. However, none of these molecular alterations is suitable either
Ming Zhao et al.
Immunity, 55(4), 623-638 (2022-04-07)
The epithelium is an integral component of mucosal barrier and host immunity. Following helminth infection, the intestinal epithelial cells secrete "alarmin" cytokines, such as interleukin-25 (IL-25) and IL-33, to initiate the type 2 immune responses for helminth expulsion and tolerance.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service