Skip to Content
Merck
All Photos(6)

Key Documents

HPA003417

Sigma-Aldrich

Anti-JAM3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-JAM-2, Anti-JAM-3, Anti-JAM-C, Anti-Junctional adhesion molecule 3, Anti-Junctional adhesion molecule C precursor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

RDSALYRCEVVARNDRKEIDEIVIELTVQVKPVTPVCRVPKAVPVGKMATLHCQESEGHPRPHYSWYRNDVPLPTDSRANPRFRNSSFHLNSETGTLVFTAVHKDDSGQYYCIASNDAGSARCEEQEMEVYDLNIG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... JAM3(83700)

General description

Junctional adhesion molecule 3 (JAM3) is a protein encoded by the JAM3 gene in humans. It is a member of the immunoglobulin superfamily. The protein is an adhesion molecule expressed by blood and lymphatic ECs, epithelial cells, fibroblasts, and circulating cells, such as monocytes, lymphocytes, and platelets.

Immunogen

Junctional adhesion molecule C precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-JAM3 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Junctional adhesion molecule C plays a role in tight junction formation, leukocyte adhesion and transendothelial migration. It plays an important role in different metastasis capacity of lymph node. It affects β1and ERK activation in human dermal lymphatic endothelial cells (HDLEC) and promotes lymphangiogenesis and nodal metastasis. It may act as a therapeutic target for preventing and treating lymphatic metastases. It has some role in the development and function of the vascular system and the brain. The gene is essential for maintaining the integrity of the cerebrovascular endothelium as well as for normal lens development in humans.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86412

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nadia A Akawi et al.
Human mutation, 34(3), 498-505 (2012-12-21)
We have recently shown that the hemorrhagic destruction of the brain, subependymal, calcification, and congenital cataracts is caused by biallelic mutations in the gene encoding junctional adhesion molecule 3 (JAM3) protein. Affected members from three new families underwent detailed clinical
Carmen Doñate et al.
Cancer research, 73(2), 640-651 (2012-12-12)
Junctional adhesion molecule C (JAM-C) is expressed by vascular endothelium and human but not mouse B lymphocytes. The level of JAM-C expression defines B-cell differentiation stages and allows the classification of marginal zone-derived (JAM-C-positive) and germinal center-derived (JAM-C-negative) B-cell lymphomas.
Bradley J Rabquer et al.
Journal of immunology (Baltimore, Md. : 1950), 185(3), 1777-1785 (2010-07-02)
Junctional adhesion molecule-C (JAM-C) is an adhesion molecule expressed by endothelial cells (ECs) that plays a role in tight junction formation, leukocyte adhesion, and transendothelial migration. In the current study, we investigated whether JAM-C is found in soluble form and
Mirko Manetti et al.
Arthritis and rheumatism, 65(1), 247-257 (2012-09-25)
Systemic sclerosis (SSc) is characterized by early perivascular inflammation, microvascular endothelial cell (MVEC) activation/damage, and defective angiogenesis. Junctional adhesion molecules (JAMs) regulate leukocyte recruitment to sites of inflammation and ischemia-reperfusion injury, vascular permeability, and angiogenesis. This study was undertaken to
Ganeshwaran H Mochida et al.
American journal of human genetics, 87(6), 882-889 (2010-11-27)
The tight junction, or zonula occludens, is a specialized cell-cell junction that regulates epithelial and endothelial permeability, and it is an essential component of the blood-brain barrier in the cerebrovascular endothelium. In addition to functioning as a diffusion barrier, tight

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service