Skip to Content
Merck
All Photos(5)

Key Documents

HPA003412

Sigma-Aldrich

Anti-PLAT antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Alteplase, Anti-Reteplase, Anti-Tissue-type plasminogen activator precursor, Anti-t- PA, Anti-t-plasminogen activator, Anti-tPA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

KYSSEFCSTPACSEGNSDCYFGNGSAYRGTHSLTESGASCLPWNSMILIGKVYTAQNPSAQALGLGKHNYCRNPDGDAKPWCHVLKNRRLTWEYCDVPSCSTCGLRQYSQPQFRIKGGLF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PLAT(5327)

General description

The gene PLAT or TPA (Tissue Plasminogen Activator) has been mapped to human chromosome 8p12.

Immunogen

Tissue-type plasminogen activator precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-PLAT antibody produced in rabbit has been used for global protein profiling to find new molecular biomarkers for common, multifactorial disorders. Anti-PLAT antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

Tissue plasminogen activator (tPA or PLAT) is a serine protease found on endothelial cells and is involved in the breakdown of blood clots. It plays an important role in synaptic regulation, plasticity and in neurogenesis. The abnormal function of this gene may be related to pathogenesis of schizophrenia. It is used for the treatment of acute ischemic stroke. An increase in endogenous levels of TPA protein has been observed in preclinical atherosclerosis, which may serve as a marker for risk of future myocardial infarction.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST85197

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silvia Hoirisch-Clapauch et al.
Seminars in thrombosis and hemostasis, 39(8), 950-954 (2013-10-11)
Schizophrenia, a disabling mental disorder, is characterized by brain atrophy, especially in the superior temporal gyrus and the medial temporal lobe, which includes the hippocampus and the amygdala. The model that better explains brain atrophy includes a trigger and a
P M Ridker et al.
Lancet (London, England), 341(8854), 1165-1168 (1993-05-08)
Endogenous tissue-type plasminogen activator (tPA) has been hypothesised to be a marker of baseline fibrinolytic capacity. We therefore tested whether tPA antigen is associated with the occurrence of future myocardial infarction (MI) among apparently healthy individuals. tPA antigen concentrations were
J G van der Bom et al.
Circulation, 95(12), 2623-2627 (1997-06-17)
Impaired fibrinolytic capacity, as assessed by euglobulin clot lysis time or plasma concentration of fibrinolytic parameters, has been associated with an increased risk of myocardial infarction (MI). We studied the association of a polymorphism in the gene for TPA and
Elisabeth B Marsh et al.
Medicine, 92(6), 317-323 (2013-10-23)
Symptomatic intracranial hemorrhage (sICH) is a known complication following administration of intravenous tissue plasminogen activator (IV tPA) for acute ischemic stroke. sICH results in high rates of death or long-term disability. Our ability to predict its occurrence is important in
Xuanbin Wang et al.
International journal of molecular sciences, 17(4), 577-577 (2016-04-20)
Hepatocellular carcinoma (HCC) is the second leading cause of cancer-related death and its prognosis remains poor due to the high risk of tumor recurrence and metastasis. Berberine (BBR) is a natural compound derived from some medicinal plants, and accumulating evidence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service