Skip to Content
Merck
All Photos(9)

Key Documents

HPA003387

Sigma-Aldrich

Anti-CDK1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CDC2, Anti-Cell division control protein 2 homolog, Anti-Cyclin-dependent kinase 1, Anti-p34 protein kinase

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, human, rat

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

LGSARYSTPVDIWSIGTIFAELATKKPLFHGDSEIDQLFRIFRALGTPNNEVWPEVESLQDYKNTFPKWKPGSLASHVKNLDENGLDLLSKMLIYDPAKRISGKMALNHPYFNDLDNQIKKM

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDC2(983)

Immunogen

Cell division control protein 2 homolog recombinant protein epitope signature tag (PrEST)

Application

Anti-CDK1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

CDK1 (cyclin-dependent kinase 1) gene encodes a protein that belongs to the Ser/Thr protein kinase family. CDKs catalyze the transfer of phosphate from ATP to specific protein substrates and function in cell cycle processes. It forms a subunit of a protein kinase complex known as M-phase promoting factor (MPF). The complex is involved in G1/S and G2/M phase transitions of eukaryotic cell cycle. In the absence of interphase Cdks (Cdk2, Cdk3, Cdk4 and Cdk6), Cdk1 binds to all cyclins, phosphorylates the retinoblastoma protein pRb and regulates the expression of genes controlled by E2F transcription factors. It drives all the events required for mammalian cell division even in the absence of other Cdks. It functions in double-strand break (DSB)-induced homologous recombination at any stage of the cell cycle. It recruits single-stranded DNA-binding complex, RPA, and the Rad51 recombination protein at the DSB ends and ensures efficient 5′ to 3′ resection.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86431

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Grzegorz Ira et al.
Nature, 431(7011), 1011-1017 (2004-10-22)
A single double-strand break (DSB) induced by HO endonuclease triggers both repair by homologous recombination and activation of the Mec1-dependent DNA damage checkpoint in budding yeast. Here we report that DNA damage checkpoint activation by a DSB requires the cyclin-dependent
Commercial Cdk1 antibodies recognize the centrosomal protein Cep152.
Gražvydas Lukinavičius et al.
BioTechniques, 55(3), 111-114 (2013-10-19)
Jeffrey A Ubersax et al.
Nature, 425(6960), 859-864 (2003-10-24)
The events of cell reproduction are governed by oscillations in the activities of cyclin-dependent kinases (Cdks). Cdks control the cell cycle by catalysing the transfer of phosphate from ATP to specific protein substrates. Despite their importance in cell-cycle control, few
Zhuo Shao et al.
Oncogenesis, 11(1), 44-44 (2022-08-05)
Pancreatic cancer has an extremely terrible prognosis and is a common cause of cancer death. In this study, the clinic value, biological function and underlying mechanisms of Zinc finger protein 655 (ZNF655) in human pancreatic cancer were evaluated. The expression
Yusuke Goto et al.
British journal of cancer, 117(3), 409-420 (2017-06-24)
Despite recent advancements, metastatic castration-resistant prostate cancer (CRPC) is not considered curative. Novel approaches for identification of therapeutic targets of CRPC are needed. Next-generation sequencing revealed 945-1248 miRNAs from each lethal mCRPC sample. We constructed miRNA expression signatures of CRPC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service