Skip to Content
Merck
All Photos(7)

Key Documents

HPA003324

Sigma-Aldrich

Anti-SLC16A1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-MCT 1, Anti-Monocarboxylate transporter 1, Anti-Solute carrier family 16 member 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

PTKAGKDKSKASLEKAGKSGVKKDLHDANTDLIGRHPKQEKRSVFQTINQFLDLTLFTHRGF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC16A1(6566)

General description

Solute carrier family 16, member 1 (SLC16A1) gene spanning 44kb is mapped to human chromosome 1p13.2-p12. The encoded protein MCT1, belongs to the monocarboxylate transporter (MCT) family.

Immunogen

Monocarboxylate transporter 1 recombinant protein epitope signature tag (PrEST)

Application

Anti-SLC16A1 antibody produced in rabbit has been used in immunostaining and immunoblotting.
Anti-SLC16A1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Solute carrier family 16, member 1(SLC16A1) gene encodes a proton-coupled monocarboxylate transporter that catalyzes the transport of many monocarboxylates such as lactate, pyruvate, branched-chain oxo acids, acetoacetate, β-hydroxybutyrate and acetate.The protein is essential for normal assimilation of nutrients. It is coexpressed with lactate dehydrogenase B (LDHB) in basal-like breast cancer. It is found to be expressed on tumor cells. It serves as a potential therapeutic target for high-risk neuroblastomas. Mutation in SLC16A1 causes profound ketoacidosis characterized with imbalanced hepatic production and extrahepatic utilization of ketone bodies.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86072

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genetic variations in the MCT1 (SLC16A1) gene in the Chinese population of Singapore
Lean CB and Lee EJD
Drug Metabolism and Pharmacokinetics, 24(5), 469-474 (2009)
Peter M van Hasselt et al.
The New England journal of medicine, 371(20), 1900-1907 (2014-11-13)
Ketoacidosis is a potentially lethal condition caused by the imbalance between hepatic production and extrahepatic utilization of ketone bodies. We performed exome sequencing in a patient with recurrent, severe ketoacidosis and identified a homozygous frameshift mutation in the gene encoding
Jun Fang et al.
Molecular pharmacology, 70(6), 2108-2115 (2006-09-27)
Neuroblastomas produce high amounts of lactic acid and upregulate the H(+)-linked monocarboxylate transporter isoform 1 (MCT1/SLC16A1). We found elevated MCT1 mRNA levels in fresh neuroblastoma biopsy samples that correlated positively with risk of fatal disease and amplification of the "proto-oncogenic"
I Tamai et al.
Biochemical and biophysical research communications, 214(2), 482-489 (1995-09-14)
A molecular mechanism for the intestinal monocarboxylic acid transport was characterized by using a proton/monocarboxylate transporter, MCT1, in Chinese hamster ovary (CHO) cells, first found by Garcia et al. (Cell, 76, 865-873, 1994). Northern blotting analysis showed that MCT1-isomers exist
MDA-MB-231 breast cancer cells fuel osteoclast metabolism and activity: A new rationale for the pathogenesis of osteolytic bone metastases
Lemma S, et al.
Biochimica et Biophysica Acta (BBA)-Molecular Basis of Disease, 1863(12), 3254-3264 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service