Skip to Content
Merck
All Photos(1)

Key Documents

HPA002740

Sigma-Aldrich

Anti-TG antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Thyroglobulin precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

KMCSADYADLLQTFQVFILDELTARGFCQIQVKTFGTLVSIPVCNNSSVQVGCLTRERLGVNVTWKSRLEDIPVASLPDLHDIERALVGKDLLGRFTDLIQSGSFQLHLDSKTFPAETIRFLQGDHFGTSPRTWFGCSEGFYQVLTSEASQDGLGCVKCPEGS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TG(7038)

Immunogen

Thyroglobulin precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Thyroglobulin (TG) is a monomeric glycoprotein produced in thyroid gland with a molecular weight 660,000. It is secreted from the endoplasmatic reticulum site. It plays a major role in the synthesis of thyroid hormone (T4, T3) and iodothyronine. It has reported that monomeric structure contains the thyroid hormones synthesis zone at both ends and glycosylation sites are scattered along the polypeptide chain of the protein. Mutations in the TG gene lead to the congenital hypothyroidism (CH).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST84470

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hakan Cangul et al.
Endocrine, 45(2), 206-212 (2013-08-21)
Mutations in the thyroglobulin (TG) gene have been reported to cause congenital hypothyroidism (CH) and we have been investigating the genetic architecture of CH in a large cohort of consanguineous/multi-case families. Our aim in this study was to determine the
T Ieiri
Nihon rinsho. Japanese journal of clinical medicine, 52(4), 869-874 (1994-04-01)
The thyroglobulin (Tg) gene is a 300-kilobase (kb) single copy gene, containing at least 42 exons, mapped in man to chromosome 8 (8q24) and codes for a glycoprotein with a molecular weight 660,000, which functions as a matrix for the
S A van de Graaf et al.
European journal of endocrinology, 136(5), 508-515 (1997-05-01)
We developed a transient transfection system for human thyroglobulin (TG) cDNA in both human thyroid cells and in COS-1 cells. Four overlapping TG cDNA fragments were amplified by reverse transcription-PCR from RNA of normal thyroid tissue. The most 5' fragment
Y Malthiéry et al.
European journal of biochemistry, 165(3), 491-498 (1987-06-15)
The mRNA encoding human thyroglobulin has been cloned and sequenced. It is made up of a 8301-nucleotide segment encoding a preprotein monomer of 2767 amino acids, flanked by non-coding 5' and 3' regions of 41 and 106 nucleotides, respectively. This

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service