Skip to Content
Merck
All Photos(5)

Documents

HPA002350

Sigma-Aldrich

Anti-C1QA antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Complement C1q subcomponent subunit A precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

GSPGNIKDQPRPAFSAIRRNPPMGGNVVIFDTVITNQEEPYQNHSGRFVCTVPGYYYFTFQVLSQWEICLSIVSSSRGQVRRSLGFCDTTNKGLFQVVSGGMVLQLQQGDQVWVEKDPKKGHIYQGSEADSVFS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... C1QA(712)

General description

C1QA (complement component 1, q subcomponent, A chain) is a versatile recognition protein with a molecular mass of 460kDa. It is synthesized by monocyte/macrophage lineage, microglia cells. Crystal structure studies have suggested a compact spherical heterotrimeric assembly held by the non-polar interactions, with a Ca2+ ion bound at the top. The bouquet type is composed of six heterotrimeric collagen-like triple helices. Its N-terminal end forms a “stalk” whereas C-terminal heterotrimeric globular domain forms individual “stems”.

Immunogen

Complement C1q subcomponent subunit A precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

C1QA (complement component 1, q subcomponent, A chain) is involved in the activation of the classical complement pathway by binding to the immune and non-immune ligands. C1 subunit of C1Q is associated with innate immunity i.e. activation of the complement classical pathway. Deficiency of C1q leads to the lupus erythematosus and glomerulonephritis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST78244

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A R Korotzer et al.
Experimental neurology, 134(2), 214-221 (1995-08-01)
Senile plaques, the pathological hallmark of Alzheimer's disease (AD), are associated with complement components, including C1q. Reactive microglia appear to be involved in the later stages of plaque development. Since tissue macrophages are known to synthesize C1q, cultured rat microglia
S Uwatoko et al.
Clinical immunology and immunopathology, 30(1), 104-116 (1984-01-01)
The molecular size of C1q-binding immunoglobulin (Ig) G complexes in systemic lupus erythematosus (SLE) sera was studied by gel filtration using C1q solid-phase radioimmunoassay (C1q SPRIA). All 15 SLE sera tested contained predominantly small-sized IgG complexes, cofractionating with monomeric IgG.
Christine Gaboriaud et al.
The Journal of biological chemistry, 278(47), 46974-46982 (2003-09-10)
C1q is a versatile recognition protein that binds to an amazing variety of immune and non-immune ligands and triggers activation of the classical pathway of complement. The crystal structure of the C1q globular domain responsible for its recognition properties has
Shui-Yi Hu et al.
BMC immunology, 14, 42-42 (2013-09-24)
Anti-glomerular basement membrane (GBM) disease is a well-known antibody-induced autoimmune disease. A few patients have glomerular C1q deposition, but it is usually absent on renal histopathology. The role of C1q deposition in kidney injury is unclear. Recently, anti-C1q antibodies are
Guobao Chen et al.
The Journal of biological chemistry, 286(40), 34941-34950 (2011-08-25)
Hereditary homozygous C1q deficiency is rare, but it almost certainly causes systemic lupus erythematosus. On the other hand, C1q levels can decline in systemic lupus erythematosus patients without apparent C1q gene defects and the versatility in C1q production is a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service