Skip to Content
Merck
All Photos(4)

Key Documents

HPA002237

Sigma-Aldrich

Anti-HS3ST1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Heparan sulfate 3-O-sulfotransferase 1 antibody produced in rabbit, Anti-Heparan sulfate D-glucosaminyl 3-O-sulfotransferase 1 antibody produced in rabbit, Anti-Heparan sulfate glucosamine 3-O-sulfotransferase 1 precursor antibody produced in rabbit, Anti-h3-OST-1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

TQVFYNHMQKHKPYPSIEEFLVRDGRLNVDYKALNRSLYHVHMQNWLRFFPLRHIHIVDGDRLIRDPFPEIQKVERFLKLSPQINASNFYFNKTKGFYCLRDSGRDRCLHESKGRAHPQVDPKLLNKLHEYFHEPNKKFFELVGR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HS3ST1(9957)

General description

HSTS3T1 (heparan sulfate-glucosamine 3-sulfotransferase 1), a type II integral membrane protein, is predominantly expressed in kidney, brain and to lesser extent in heart and lung. It is composed of an amino-terminal region and a homologous carboxyl-terminal sulfotransferase domain.

Immunogen

Heparan sulfate glucosamine 3-O-sulfotransferase 1 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

HS3ST1 (heparan sulfate-glucosamine 3-sulfotransferase 1) plays a major role in the rate limiting step of the synthesis of anticoagulant heparan sulfate. It converts the nonanticoagulant heparan sulfate to anticoagulant heparan sulfate by transferring sulfate group from adenosine 3′-phosphophate 5′-phospho-sulfate to the 3-OH position of glucosamine.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86548

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N W Shworak et al.
The Journal of biological chemistry, 272(44), 28008-28019 (1997-11-05)
The cellular rate of anticoagulant heparan sulfate proteoglycan (HSPGact) generation is determined by the level of a kinetically limiting microsomal activity, HSact conversion activity, which is predominantly composed of the long sought heparan sulfate D-glucosaminyl 3-O-sulfotransferase (3-OST) (Shworak, N. W.
J Liu et al.
The Journal of biological chemistry, 274(8), 5185-5192 (1999-02-13)
The 3-O-sulfation of glucosamine residues is an important modification during the biosynthesis of heparan sulfate (HS). Our previous studies have led us to purify and molecularly clone the heparan sulfate D-glucosaminyl 3-O-sulfotransferase (3-OST-1), which is the key enzyme converting nonanticoagulant
N W Shworak et al.
The Journal of biological chemistry, 274(8), 5170-5184 (1999-02-13)
3-O-Sulfated glucosaminyl residues are rare constituents of heparan sulfate and are essential for the activity of anticoagulant heparan sulfate. Cellular production of the critical active structure is controlled by the rate-limiting enzyme, heparan sulfate D-glucosaminyl 3-O-sulfotransferase-1 (3-OST-1) (EC 2.8.2.23). We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service