H6771
2-Hydroxytetradecanoic acid
≥98% (capillary GC)
Synonym(s):
2-Hydroxymyristic acid, DL-α-Hydroxymyristic acid
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C14H28O3
CAS Number:
Molecular Weight:
244.37
Beilstein:
1725369
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77
Recommended Products
Assay
≥98% (capillary GC)
lipid type
saturated FAs
storage temp.
2-8°C
SMILES string
CCCCCCCCCCCCC(O)C(O)=O
InChI
1S/C14H28O3/c1-2-3-4-5-6-7-8-9-10-11-12-13(15)14(16)17/h13,15H,2-12H2,1H3,(H,16,17)
InChI key
JYZJYKOZGGEXSX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
An analog of myristic acid that becomes metabolically activated in cells to form 2-hydroxymyristoyl-CoA. Inhibitor of protein myristoylation.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
M S Trent et al.
The Journal of biological chemistry, 276(46), 43132-43144 (2001-09-06)
Polymyxin-resistant mutants of Escherichia coli and Salmonella typhimurium accumulate a novel minor lipid that can donate 4-amino-4-deoxy-l-arabinose units (l-Ara4N) to lipid A. We now report the purification of this lipid from a pss(-) pmrA(C) mutant of E. coli and assign
S M Cordo et al.
Microbes and infection, 1(8), 609-614 (1999-12-28)
The effects of two myristic acid analogs on Junin virus (JV) replication were investigated. The compounds chosen for the study were DL-2-hydroxymyristic acid (2OHM), an inhibitor of N-myristoyltransferase (NMT), which binds the enzyme and blocks protein myristoylation, and 13-oxamyristic acid
J S Gunn et al.
Infection and immunity, 68(6), 3758-3762 (2000-05-19)
The PhoP-PhoQ two-component system is necessary for the virulence of Salmonella spp. and is responsible for regulating several modifications of the lipopolysaccharide (LPS). Mutagenesis of the transcriptional regulator phoP resulted in the identification of a mutant able to activate transcription
Abdulla S Kazim et al.
Biochemical and biophysical research communications, 491(3), 740-746 (2017-07-26)
High blood glucose triggers the release of insulin from pancreatic beta cells, but if chronic, causes cellular stress, partly due to impaired Ca
L I Sly et al.
International journal of systematic bacteriology, 47(1), 211-213 (1997-01-01)
Determination of the 16S rRNA gene sequence of Caulobacter subvibrioides ATCC 15264T (T = type strain) confirmed that this species is a member of the alpha subclass of the Proteobacteria and showed that it is phylogenetically most closely related to
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service