All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C29H50O6
CAS Number:
Molecular Weight:
494.70
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77
Recommended Products
Quality Level
application(s)
agriculture
SMILES string
CC[C@@H](C(C)C)[C@H](O)[C@@H](O)[C@@H](C)[C@H]1CC[C@H]2[C@@H]3COC(=O)[C@H]4C[C@H](O)[C@H](O)C[C@]4(C)[C@H]3CC[C@]12C
InChI
1S/C29H50O6/c1-7-17(15(2)3)26(33)25(32)16(4)19-8-9-20-18-14-35-27(34)22-12-23(30)24(31)13-29(22,6)21(18)10-11-28(19,20)5/h15-26,30-33H,7-14H2,1-6H3/t16-,17-,18-,19+,20-,21-,22+,23-,24+,25-,26-,28+,29+/m0/s1
InChI key
HJIKODJJEORHMZ-DNVPGCTHSA-N
Application
22(S),23(S)-Homobrassinolide has been used as a brassinosteroid supplement:
- to study its effects on DNA methylation, callus growth and regeneration in barley.
- to study its effects on cell elongation in Arabidopsis thaliana.
- to study its effects on cell division, root germination and anti-oxidant system enzymes in barley.
Biochem/physiol Actions
22(S),23(S)-Homobrassinolide is a brassinosteroid. It might play a role in the control of plant embryogenesis.
Exhibits plant growth regulatory activity.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
P Muthuraman et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 41(1), 1-9 (2010-06-24)
This study aimed to investigate the effect of the plant growth regulator 28-homobrassinolide (HB) on the hexokinase I (HK I) enzyme gene expression in the tissues of normal and streptozotocin-induced diabetic rats. Normal and diabetic rats were administered 50 microg
Shamsul Hayat et al.
Plant physiology and biochemistry : PPB, 53, 61-68 (2012-02-11)
Among various environmental stresses, salt stress is extensively damaging to major crops all over the world. An experiment was conducted to explore the role of exogenously applied 28 homobrassinolide (HBL) and salicylic acid (SA) on growth, photosynthetic parameters, transpiration and
Alexander Yu Misharin et al.
Steroids, 75(3), 287-294 (2010-01-26)
Toxicity of eight 22,23-dihydroxystigmastane derivatives (four pairs of (22R,23R)- and (22S,23S)-isomers differing in steroid backbone structure) to human breast carcinoma MCF-7 cells was compared. For every pair of structurally related compounds, (22R,23R) isomer was found to be significantly more toxic
Indu Sharma et al.
Indian journal of biochemistry & biophysics, 47(3), 172-177 (2010-07-27)
28-Homobrassinolide (28-HBL), a brassinosteroid is reported to play significant role in diverse physiological processes. It induces a range of cellular and adaptive responses to a range of environmental stresses. Cadmium (Cd) is a non-essential metal which alters various physiological processes
Mohammad Yusuf et al.
Chemosphere, 85(10), 1574-1584 (2011-09-09)
The objective of this study was to establish relationship between boron induced oxidative stress and antioxidant system in Vigna radiata plants and also to investigate whether brassinosteroids will enhance the level of antioxidant system that could confer tolerance to the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service