GW22778
Anti-GSK3a antibody produced in chicken
affinity isolated antibody, buffered aqueous solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
Recommended Products
biological source
chicken
Quality Level
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
species reactivity
rat, mouse, human
manufacturer/tradename
Genway 15-288-22778
technique(s)
indirect ELISA: suitable
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... GSK3A(2931)
Immunogen
Immunogen Sequence: GI # 11995474, sequence 89-100
Synthetic peptide sequence of glycogen synthase kinase 3α.
Application
Anti-GSK3α antibody produced in chicken is suitable for indirect ELISA and western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.
Biochem/physiol Actions
Glycogen synthase kinase-3α is an enzyme encoded by the GSK3A gene in humans and is located on chromosome 19q13.2. It is a serine-threonine kinase that exists as two isoforms, α and β. GSK-3α is widely expressed throughout the follicles and plays a role in cancer development such as acute myeloid leukemia (AML). It is an important component of the therapeutic actions of drugs used to treat mood disorders and can be a potential target for developing new therapeutic interventions.
Physical form
Solution in phosphate buffered saline containing 0.02% sodium azide.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
P C Shaw et al.
Genome, 41(5), 720-727 (1998-11-11)
Glycogen synthase kinase-3 (GSK-3) is a serine-threonine kinase that exists as two isoforms, alpha and beta, encoded by separate genes. Phosphorylation targets include a variety of cytoplasmic and nuclear proteins. Recent studies found that neurofilaments, amyloid precursor protein, and tau
Xiaohua Li et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 35(11), 2143-2154 (2010-07-30)
Little is known regarding the mechanisms underlying the complex etiology of mood disorders, represented mainly by major depressive disorder and bipolar disorder. The 1996 discovery that lithium inhibits glycogen synthase kinase-3 (GSK3) raised the possibility that impaired inhibition of GSK3
Koichi Yamauchi et al.
Archives of dermatological research, 302(4), 263-270 (2009-08-26)
Beta-catenin is involved in the hair follicle morphogenesis and stem cell differentiation, and inhibition of glycogen synthase kinase-3 (GSK-3) increases beta-catenin concentration in the cytoplasm. To examine the effects of GSK-3 inhibition on the hair follicle epithelium, we first examined
Versha Banerji et al.
The Journal of clinical investigation, 122(3), 935-947 (2012-02-14)
Acute myeloid leukemia (AML) is the most common form of acute leukemia in adults. Long-term survival of patients with AML has changed little over the past decade, necessitating the identification and validation of new AML targets. Integration of genomic approaches
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service