Skip to Content
Merck
All Photos(1)

Key Documents

GW21321

Sigma-Aldrich

Anti-RGS16 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-RGSR, Anti-Regulator of G-protein signaling 16

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse, rat

manufacturer/tradename

Genway 15-288-21321

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RGS16(6004)

Related Categories

Immunogen

Immunogen Sequence: GI # 4506513, sequence 130-202
Recombinant regulator of G-protein signalling 16

Application

Anti-RGS16 antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Regulator of G-protein signaling 16 is a protein encoded by the RGS16 gene in humans and is mapped to chromosome 1q25-1q31. It is found to be expressed in 17 human gastrointestinal cancer cell lines. RGS16 expression is higher in colorectal cancer (CRC) tissue when compared to normal tissue. It can act as a predictive marker for patient prognosis of CRC. Loss of RGS16 in some breast tumors enhances PI3K signaling elicited by growth factors and thereby promotes proliferation and TKI evasion downstream of HER activation.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genqing Liang et al.
The Journal of biological chemistry, 284(32), 21719-21727 (2009-06-11)
Aberrant activity of the phosphatidylinositol 3-kinase (PI3K) pathway supports growth of many tumors including those of breast, lung, and prostate. Resistance of breast cancer cells to targeted chemotherapies including tyrosine kinase inhibitors (TKI) has been linked to persistent PI3K activity
B E Snow et al.
Gene, 206(2), 247-253 (1998-04-04)
Regulators of G-protein signaling (RGS) constitute a family of GTPase-activating proteins with varying tissue-specific expression patterns and G-protein alpha subunit specificities. Here, we describe the molecular cloning of the human RGS-r/RGS16 cDNA, encoding a predicted polypeptide of 23kDa that shows
Norikatsu Miyoshi et al.
Annals of surgical oncology, 16(12), 3507-3514 (2009-09-18)
The RGS family, comprising 22 homologues of proteins, plays a role in cellular proliferation, differentiation, membrane trafficking, and embryonic development through the involvement of the mitogen-activated protein kinase signaling pathway. In order to demonstrate the importance of RGS16 expression for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service