Skip to Content
Merck
All Photos(1)

Documents

GW21255B

Sigma-Aldrich

Anti-Exportin T antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Exportin (tRNA)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.43

biological source

chicken

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

manufacturer/tradename

Genway 15-288-21255B

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... XPOT(11260)

General description

XPOT (exportin T) is a tRNA nuclear export receptor found in vertebrates. This protein shuttles between the cytoplasm and nucleus. It belongs to the importin-β family, and has a distant relation to Los1p protein found in yeast.

Immunogen

Immunogen Sequence: GI # 8051636, sequence 5-234
Recombinant TRNA exportin

Application

Anti-Exportin T antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Exportin-T acts as a specific mediator of tRNA export. Exportin-T is a RanGTP-binding, importin β-related factor with predominantly nuclear localization that shuttles rapidly between nucleus and cytoplasm. It interacts with nuclear pore complexes as well. Exportin-t binds tRNA directly and with high affinity. It is responsible for majority of tRNA export in cells and plays an essential role in the transport of tRNA. It is associated with intracellular tRNA trafficking and is in turn associated with discrete changes in the composition of cellular cytoplasmic multi-protein complexes containing tRNA.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takuji Suzuki et al.
Biomedical research (Tokyo, Japan), 27(2), 89-92 (2006-05-19)
The active form of vitamin D3, 1alpha,25-dihydroxyvitamin D3 (DVD), is a potent inducer of cell differentiation and inhibits proliferation of cells such as human promyelocytic leukemia HL-60 cells. In the present study, we examined the effects of DVD on the
U Kutay et al.
Molecular cell, 1(3), 359-369 (1998-07-14)
In eukaryotes, tRNAs are synthesized in the nucleus and after several maturation steps exported to the cytoplasm. Here, we identify exportin-t as a specific mediator of tRNA export. It is a RanGTP-binding, importin beta-related factor with predominantly nuclear localization. It
G J Arts et al.
The EMBO journal, 17(24), 7430-7441 (1998-12-19)
Exportin-t (Xpo-t) is a vertebrate nuclear export receptor for tRNAs that binds tRNA cooperatively with GTP-loaded Ran. Xpo-t antibodies are shown to efficiently block tRNA export from Xenopus oocyte nuclei suggesting that it is responsible for at least the majority
C Kruse et al.
The Biochemical journal, 346 Pt 1, 107-115 (2000-02-05)
Vigilin is a ubiquitous multi heterogeneous nuclear ribonucleoprotein (hnRNP) K homologous (KH)-domain protein. Here we demonstrate that purified recombinant human vigilin binds tRNA molecules with high affinity, although with limited specificity. Nuclear microinjection experiments revealed for the first time that
G J Arts et al.
Current biology : CB, 8(6), 305-314 (1998-03-25)
Transport of macromolecules between the nucleus and cytoplasm of eukaryotic cells is mediated by nuclear import and export receptors. The receptors identified to date are members of a family of Ran GTPase-binding proteins whose founding member is importin-beta. Interaction between

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service