Skip to Content
Merck
All Photos(1)

Key Documents

GW21030

Sigma-Aldrich

Anti-Interleukin-8 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CXCL8, Anti-IL-8

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ELISA (i)
WB
clone:
polyclonal
species reactivity:
human
citations:
4
technique(s):
indirect ELISA: suitable
western blot: suitable

biological source

chicken

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

manufacturer/tradename

Genway 15-288-21030

technique(s)

indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IL8(3576)

Related Categories

General description

IL-8 (interleukin-8), also called CXCL8, is an α chemokine belonging to the cysteine-x-cysteine (CXC) chemokine subfamily. It is a pro-inflammatory cytokine and is secreted by multiple types of tissues and blood cells such as, monocytes, neutrophils, tissue macrophages, T-lymphocytes, endothelial and epithelial cells, and fibroblasts. It is pro-angiogenic and tumorigenic, and is produced by cancer cells such as, colorectal cancer cells.

Immunogen

Immunogen Sequence: GI # 124359, sequence 28-99
Recombinant interleukin-8

Application

Anti-Interleukin-8 antibody produced in chicken is suitable for indirect ELISA and western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Interleukin-8 (IL 8) is a proinflammatory chemokine that activates multiple signaling pathways. The exosomes released from chronic myelogenous leukemia (CML) cells stimulate bone marrow stromal cells to produce IL 8. It plays a very important regulatory function in tumorigenesis. Its polymorphism is associated with a significantly increased risk of cancers and may be useful for cancer susceptibility studies.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Bălăşoiu et al.
Romanian journal of morphology and embryology = Revue roumaine de morphologie et embryologie, 55(2 Suppl), 575-578 (2014-09-03)
Colorectal cancer represents a major cause of mortality and morbidity and occupies the third place as cancer incidence and the fourth place as cancer mortality. Colorectal cancer causes 608 000 deaths per year, despite correct applied treatment (surgery and chemotherapy).
Kai Yang et al.
International journal of molecular sciences, 15(11), 21286-21298 (2014-11-20)
The objective of this study was to analyze the expression levels of IL-8 in serum and liver tissues from patients with chronic hepatitis B (CHB) infection and to investigate whether IL-8 may antagonize interferon-alpha (IFN-α) antiviral activity against HBV. IL-8
Chiara Corrado et al.
Cancer letters, 348(1-2), 71-76 (2014-03-25)
Chronic myelogenous leukemia (CML) is a myeloproliferative disorder characterized by the Bcr-Abl oncoprotein with constitutive tyrosine kinase activity. Exosomes are nanovesicles released by cancer cells that are involved in cell-to-cell communication thus potentially affecting cancer progression. It is well known
Ziliang Wang et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 35(7), 7115-7123 (2014-04-25)
Emerging evidences show that interleukin-8 (IL-8) has important regulatory functions in tumorigenesis. IL-8 -251A/T is a single nucleotide polymorphism in the promoter region of the IL-8 gene and affects IL-8 production. Analysis of previous studies on the association of -251A/T

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service