Skip to Content
Merck
All Photos(1)

Key Documents

GW20008F

Sigma-Aldrich

Anti-α-2-Macroglobulin antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-A2MG

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse, rat

manufacturer/tradename

Genway 15-288-20008F

technique(s)

indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... A2M(2)

Immunogen

α-2-Macroglobulin (validated) native human protein.
Immunogen Sequence: GI # 68765, sequence 1-1474

Application

Anti-α-2-Macroglobulin antibody produced in chicken is suitable for western blotting at a working dilution of 1:500 and for cell staining at a working dilution of 1:200. It is also used for indirect ELISA.

Biochem/physiol Actions

A2M (α-2-macroglobulin) inhibits all four classes of proteinases by a unique ′trapping′ mechanism. This protein undergoes a conformational change when a proteinase binds and cleaves a specific peptide stretch called the "bait region" in the protein. This modified protein traps the bound proteinase It is involved in the clearance and degradation of Aβ, which is a major component of beta-amyloid deposits. Therefore, it is associated with Alzheimer′s disease (AD).

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Hall et al.
FEBS letters, 254(1-2), 111-114 (1989-08-28)
Alpha 2-Macroglobulin (alpha 2 M) was isolated from plasma of the freshwater crayfish, Pacifastacus leniusculus, using ultracentrifugation, ion-exchange chromatography and gel filtration techniques. The Pacifastacus alpha 2 M molecule (P alpha 2 M) was radio-actively labeled in the thiol ester
X Huang et al.
The Journal of nutrition, 130(5S Suppl), 1488S-1492S (2000-05-10)
Alzheimer's disease (AD) is characterized by amyloid deposits within the neocortical parenchyma and the cerebrovasculature. The main component of these predominantly extracellular collections, Abeta, which is normally a soluble component of all biological fluids, is cleaved out of a ubiquitously
S L Gonias et al.
Biochimica et biophysica acta, 705(3), 306-314 (1982-08-10)
Circular dichroism spectroscopy, sedimentation velocity and ultraviolet difference spectroscopy were used to compare alpha 2-macroglobulin, alpha 2-macroglobulin-trypsin complex and alpha 2-macroglobulin-methylamine complex. The circular dichroic spectrum of native alpha 2-macroglobulin is significantly changed in shape and magnitude following reaction with
M P Cuajungco et al.
Annals of the New York Academy of Sciences, 920, 292-304 (2001-02-24)
Alzheimer's disease is a rapidly worsening public health problem. The current lack of effective treatments for Alzheimer's disease makes it imperative to find new pharmacotherapies. At present, the treatment of symptoms includes use of acetylcholinesterase inhibitors, which enhance acetylcholine levels
Vivian Lobo et al.
Molecular human reproduction, 25(3), 137-155 (2018-12-28)
Glucose regulated protein 78 (GRP78) is expressed on cell surface in exceptional conditions as seen in cancer cells and macrophages. We have reported its membrane localization in sperm. The functional significance of its surface localization in sperm is an enigma.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service