GE65152105050250
SpeedBeads magnetic carboxylate modified particles
Cytiva 65152105050250, 0.70-1.10 μm avg. part. size, suspension (5% Solids)
Synonym(s):
SpeedBeads
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.56
Recommended Products
ligand
(carboxylate modified particles)
form
suspension (5% Solids)
packaging
pkg of 15 mL
manufacturer/tradename
Cytiva 65152105050250
concentration
50 mg/mL
avg. part. size
0.70-1.10 μm
shipped in
wet ice
storage temp.
2-8°C
Application
Sera-Mag™ SpeedBeads and Sera-Mag™ Carboxylate-Modified Magnetic Particles typical application uses include sample preparation, proteomics, nucleic acid isolation, and immunoassay applications.
Features and Benefits
Sera-Mag™ SpeedBeads and Sera-Mag™ Carboxylate-Modified Magnetic Particles allow convenient covalent coupling of target molecules.
- Carboxylic groups on the surface permit easy covalent coupling to biomolecules of interest using convenient carbodiimide chemistry.
- Different levels of hydrophobicity/hydrophilicity available.
- Available in Sera-Mag™ SpeedBeads and original Sera-Mag™ versions.
- Excellent sensitivity and low nonspecific binding for greater accuracy.
- Fast Magnetic Response Times and Stable Physical Integrity
Analysis Note
To view the Certificate of Analysis for this product, please visit www.cytiva.com.
Legal Information
Sera-Mag is a trademark of Cytiva
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Structural variation detection by proximity ligation from FFPE tumor tissue.
Troll C, et al.
bioRxiv, 266023-266023 (2018)
Design and specificity of long ssDNA donors for CRISPR-based knock-in.
Li H, et al.
bioRxiv, 178905-178905 null
Magdalena Surman et al.
International journal of molecular sciences, 21(8) (2020-04-26)
Cutaneous melanoma (CM) is an aggressive type of skin cancer for which effective biomarkers are still needed. Recently, the protein content of extracellular vesicles (ectosomes and exosomes) became increasingly investigated in terms of its functional role in CM and as
Jan Leipert et al.
Lab on a chip, 19(20), 3490-3498 (2019-09-19)
While LC-MS-based proteomics with high nanograms to micrograms of total protein has become routine, the analysis of samples derived from low cell numbers is challenged by factors such as sample losses, or difficulties encountered with the manual manipulation of small
G Stefano Brigidi et al.
Cell, 179(2), 373-391 (2019-10-05)
Cells regulate gene expression in response to salient external stimuli. In neurons, depolarization leads to the expression of inducible transcription factors (ITFs) that direct subsequent gene regulation. Depolarization encodes both a neuron's action potential (AP) output and synaptic inputs, via
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service