Skip to Content
Merck
All Photos(1)

Key Documents

GE29-0510-21

HisTrap High Performance

Cytiva 29-0510-21, pack of 1 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151816
NACRES:
NA.56

packaging

pack of 1 mL

manufacturer/tradename

Cytiva 29-0510-21

availability

not available in North America

parameter

flow rate
4 ml/min (1 ml), 5 ml/min (5ml) flow rate (H2O at room temperature)
42 psi (H2O at room temperature.)

bed size

7 mm × 25 mm

bed volume

1 mL

column I.D.

7 mm

matrix

highly cross-linked 6% agarose

avg. part. size

34 μm

cleaning

2-14(Ni2+-stripped medium.)

working range

3-12(Ni2+-stripped medium)

capacity

≥40 mg binding capacity (histidine-tagged protein/ml medium)(H2O at room temperature.)
≥40 mg binding capacity (histidine-tagged protein/ml medium)(Protein binding capacity is protein-to-protein dependent.)

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

HisTrap HP columns are prepacked with Ni Sepharose High Performance and designed for simple, high-resolution purification of histidine-tagged proteins by immobilized metal ion affinity chromatography (IMAC).

Application

HisTrap HP columns are designed for simple, one-step purification of histidine-tagged proteins. The columns are prepacked with Ni Sepharose High Performance, which has high binding capacity and low nickel ion leakage that ensures reliable capture of target protein in repeated IMAC purifications.

Features and Benefits

  • High binding capacity, at least 40 mg/ml chromatography medium.
  • Compatible with a wide range of reducing agents, detergents, denaturants, and other additives.
  • Negligible Ni2+ leakage
  • Simple manual operation with a syringe, pump, or chromatography system such as AKTA design

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Legal Information

HisTrap is a trademark of Cytiva

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service