Skip to Content
Merck
All Photos(2)

Key Documents

GE29-0491-04

HiTrap® MabSelect SuRe

Cytiva 29-0491-04, pack of 1 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151816
NACRES:
NA.56

ligand

alkali-stabilized protein A-derived (E. coli)

packaging

pack of 1 mL

manufacturer/tradename

Cytiva 29-0491-04

availability

not available in North America

storage condition

(20% Ethanol)
(20% Ethanol, 0.1 M Potassium Phosphate buffer, pH 8.0)

parameter

42 psi

bed size

7 mm × 25 mm

bed volume

1 mL

column I.D.

7 mm

matrix

highly cross-linked agarose

avg. part. size

85 μm (d₅₀v is the median particle size of the cumulative volume distribution)

cleaning

2-13.7

working range

3-12

capacity

≥30 mg binding capacity (humanlgG/ml at 2.4 min residence time)

suitability

suitable for bioprocess medium

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Determined at 10% breakthrough by frontal analysis at a mobile phase velocity of 500 cm/h in a column with a bed height of 20 cm.
HiTrap MabSelect SuRe columns are prepacked with MabSelect SuRe, a BioProcess medium for purification of monoclonal antibodies from large sample volumes. The novel alkali-tolerant recombinant Protein A ligand allows cleaning-in-place (CIP).
pH below 3 is sometimes required to elute strongly bound IgG species. However, protein ligands may hydrolyze at very low pH.

Features and Benefits

  • Withstands rigorous CIP and sanitization procedures with 0.1 to 0.5 M NaOH.
  • Novel ligand design gives enhanced protease resistance resulting in lower ligand leakage.
  • Oriented coupling of recombinant Protein A to the matrix via an engineered C-terminal cysteine enhances IgG binding capacity.
  • Fewer regulatory concerns due to the total absence of mammalian culture in the ligand production and purification.
  • Prepacked HiTrap columns for process development, screening of purification conditions, and small-scale purification of Mabs.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Other Notes

The pressure over the packed bed varies depending on a range of parameters such as the characteristics of the chromatography medium and the column tubing used.

Legal Information

Free MabSelect SuRe Ligand
"MabSelect SuRe Ligand Restricted License" and "Cys-rProtein A Ligand Restricted License" are protected by the following patents and equivalent patents and patent applications in other countries: US 6,399,750 and EP 1123389. A free, non-transferable limited license to use this product for internal analytical purposes only accompanies the purchase of the product from a Cytiva company and its licensed distributors. Any other use will require a separate license from a Cytiva company.
HiTrap is a registered trademark of Cytiva
MabSelect SuRe is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service