Key Documents
GE29-0230-51
Frac30 fraction collector
round
About This Item
UNSPSC Code:
41105300
NACRES:
NA.55
material
round
manufacturer/tradename
Cytiva 29-0230-51
Related Categories
General description
Frac30 fraction collector is controlled via the ÄKTA start touch screen instrument display or using UNICORN start software on a computer and 30 allows you to collect up to 30 fractions and supports four tube sizes, ranging from 1.5 mL to 15 mL.
Fractions can be collected in volumes ranging from 0.5 mL to 15 mL. The entire fraction collector rack can be transferred as required.
available in Europe only