Skip to Content
Merck
All Photos(1)

Key Documents

GE28-9909-46

Sigma-Aldrich

Superdex® 200 Increase 3.2/300

column L × I.D. 30 cm × 3.2 mm, 8.6 μm particle size

Synonym(s):

Superdex 200 Increase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

material

glass column
titanium frit

product line

Superdex®

mol wt

10000-600000 Da (globular proteins)

packaging

1 ea of

parameter

<0.15 ml/min flow rate
3.0 Mpa (652 psi)
4-50 mL sample volume

bed volume

~2.4 mL

column L × I.D.

30 cm × 3.2 mm

matrix

agarose-dextran composite
polymeric

particle size

8.6 μm

cleaning

1-14

working range

3-12

separation technique

size exclusion (SEC)

General description

Superdex™ 200 Increase size exclusion chromatography columns are well suited for high-resolution analysis and small-scale purification of monoclonal antibodies (mAb) and other biomolecules with Mr~ 10 000 to ~ 600 000.

Superdex™ 200 Increase is a next-generation, dextran-agarose composite matrix for SEC. With smaller and more rigid beads than their predecessors, Superdex™ Increase columns deliver higher resolution purification in shorter run times.

Superdex™ 200 Increase has a selectivity curve optimized for excellent resolution of antibodies and macromolecules within the range Mr ~ 100 000 to ~ 300 000. It is well suited for detection and separation of antibody monomers, dimers, and aggregates present in monoclonal antibody preparations. The outstanding resolving power of this resin facilitates protein characterization, including analysis of antibody size variants, assessment of membrane protein size homogeneity, and the study of protein-protein interactions.

Superdex™ 200 Increase is optimized to give high resolution separation for proteins with molecular weights in the range Mr ~ 100 000 to ~ 300 000. Superose™ 6 Increase provides a complementary fractionation range, resolving very large protein complexes and macromolecules that Superdex 200 Increase cannot separate.

This agarose-based resin is alkali-resistant and supports cleaning-in-place (CIP) procedures. This capability allows the same column to be used for different proteins, with minimal risk of carry-over between samples.

Features and Benefits

  • Enhanced performance: improved resolution and runtime compared to predecessor Superdex™ 200.
  • Very high resolution: small bead size and narrow particle size distribution provide high resolution, for high protein purity.
  • High flow rates: rigid beads give excellent pressure/flow properties.

Suitability

Long term use: all commonly used aqueous buffers, up to 8 M urea, ionic and non-ionic detergents, up to 6M guanidine hydrochloride
for both preparative and analytical purposes

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Superdex is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service