GE28-9180-11
PD MidiTrap™ G-10
Cytiva 28-9180-11, pack of 50 columns
Synonym(s):
PD MidiTrap
About This Item
Recommended Products
feature
autoclavable In wet form (pH 7.0) at 120°C for 30 min
wetted part: no
packaging
pack of 50 columns
manufacturer/tradename
Cytiva 28-9180-11
parameter
flow rate (The beads behave as rigid spheres obeying Darcy′s Law.)
bed size
13 mm × 40 mm
bed volume
5.3 mL
column I.D.
13 mm
matrix
cross-linked dextran
particle size
40-120 μm (dry)
55-165 (wet)
working range
2-13
General description
Features and Benefits
- Easy cleanup of peptides, small proteins, or carbohydrates larger than Mr 700, before downstream analysis such as liquid chromatography, electrophoresis, and MS.
- Efficient removal of contaminants such as salts, dyes, and radioactive labels.
- High desalting capacity.
- Preparation of samples in the volume range 400 to 1000 μl.
Storage and Stability
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
3 - Flammable liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service